Clariom™ S Assay HT, human
Clariom™ S Assay HT, human
Applied Biosystems™

Clariom™ S Assay HT, human

Obtain a gene-level view of the human transcriptome with Clariom S Assays HT for human samples. Clariom S Assays serveRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90297096 arrays
90296924 arrays
Catalog number 902970
Price (USD)
-
Number of Arrays:
96 arrays
Ask our AI about this Product
Obtain a gene-level view of the human transcriptome with Clariom S Assays HT for human samples. Clariom S Assays serve as a next generation transcriptome-wide gene-level expression profiling tool, which allows for the fastest, simplest, and most scalable path to generating the results you need for your research. Based on industry-leading microarray technology, the novel Human HT Clariom S Assay design provides extensive coverage of all known well-annotated genes, compatibility with research sample types, scalable formats, and flexible data analysis software. Clariom S Assays HT are the tools of choice to find expression biomarkers with known function as quickly, easily, and cost-effectively as possible.

Find answers, move on
Although the number of known transcribed genes has expanded rapidly in recent years, knowledge of the function of each gene is still evolving. Many genes and transcripts found in databases are poorly annotated or unannotated, which can complicate and prolong data analysis and interpretation. Human HT Clariom S Assays focus on well-annotated genes, providing researchers with the ability to perform gene-level expression profiling studies and to quickly assess changes in key genes and pathways. With less time required for data analysis, Clariom S Assays HT for human help researchers reach conclusions more rapidly.

Simple, swift biomarker discovery
• Accurately measure gene-level expression from >20,000 well-annotated genes to get to answers quickly.
• Choose a format that suits your throughput needs, processing from 1 to 192 samples a day.
• Go from data to insight in minutes with intuitive, highly visual, free analysis software designed for the biologist.

When you have precious samples, get it right the first time
• Generate robust expression profiles from as little as 100 pg of total RNA–as few as 10 cells.
• Utilize RNA from various sample types including blood, cells, and fresh/fresh-frozen or FFPE tissues.
• Preserve sample integrity and reduce data variability with an assay that does not require a globin or rRNA removal step.
• Save time and money with fully automated sample preparation options.

Clariom S solutions are available formats for single-sample (cartridge array) processing on the GeneChip™ 3000 instrument system and high-throughput automated processing (plate array) on the GeneTitan™ HT Microarray System, offering the flexibility to accommodate both small and large cohort studies. The complete solution comes with reagents and fast, simple Transcriptome Analysis Console (TAC) software to analyze and visualize global expression patterns of genes, pathways, and network interactions in minutes.

Get the truest level of gene-level expression
To generate robust gene-level expression, Human HT Clariom S Assays detect only the exons present in all known transcript isoforms expressed from a single gene locus-constitutive exons. This differs from other gene-level array technologies and shallow RNA-Seq, which provide either a biased view of gene expression or data that are complicated by variation in expression of transcript variants. By detecting only constitutive exons throughout the length of each known gene, Human HT Clariom S Assays generate the most accurate and truest measurement of gene-level expression available today.

Keeping biomarker identification across the transcriptome simple and swift, Clariom S Assays HT for human provide you with the coverage you require, the reproducibility you need, and the insights you want to act on your discoveries.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Array Format96-array Plate
ArrayTranscriptome Profiling
Sample TypeRNA
For Use With (Application)Microarray Analysis
No. of Reactions96
Product LineApplied Biosystems™
Number of Arrays96 arrays
Product TypeS Assay HT
Quantity96 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesHuman
Unit SizeEach
Contents & Storage
• Poly-A RNA Control Kit, store at -20°C
• Hybridization Control Kit, store at -20°C
• HT WT Terminal Labeling Kit, store at -20°C
• WT Amplification Kit, Module 1, store at -20°C
• WT Amplification Kit, Module 2, store at 4°C
• Clariom S 96-Array Plate, human, store at 4°C
Have questions about this product? Ask our AI assisted search.
What reagent kit should I use with my array?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Frequently asked questions (FAQs)

Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Share catalog number, name or link

1x1 image pixel for data collection