Thermo Scientific™
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Catalog number: 942339395041
Have Questions?
Thermo Scientific™
Graphite Furnace Cuvettes for Atomic Absorption (AA) Spectrometers
Catalog number: 942339395041
Have Questions?
Discover different types of graphite furnace cuvettes suitable for any analytical application using the Thermo Scientific™ GFS33/GFS35/GFS35Z Graphite Furnace and Autosampler modules
Catalog Number
942339395041
Unit Size
Each
Description
Extended Lifetime Cuvettes
Price (USD)
Full specifications
Quantity
TypeGraphite Furnace Cuvette
DescriptionExtended Lifetime Cuvettes
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Standard Cuvettes
Price (USD)
Full specifications
Quantity
TypeGraphite Furnace Cuvette
DescriptionStandard Cuvettes
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Pyro-Coated Cuvettes
Price (USD)
Full specifications
Quantity
TypeGraphite Furnace Cuvette
DescriptionPyro-Coated Cuvettes
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Omega Cuvette with Integral Platform
Price (USD)
Full specifications
Quantity
TypeGraphite Furnace Cuvette
DescriptionOmega Cuvette with Integral Platform
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)Quantity
942339395041Full specifications
EachExtended Lifetime Cuvettes
TypeGraphite Furnace Cuvette
DescriptionExtended Lifetime Cuvettes
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
942339395031Full specifications
EachStandard Cuvettes
TypeGraphite Furnace Cuvette
DescriptionStandard Cuvettes
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
942339395071Full specifications
EachPyro-Coated Cuvettes
TypeGraphite Furnace Cuvette
DescriptionPyro-Coated Cuvettes
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
942349020101Full specifications
EachOmega Cuvette with Integral Platform
TypeGraphite Furnace Cuvette
DescriptionOmega Cuvette with Integral Platform
For Use With (Equipment)GFS33 or GFS35 Furnaces
Unit SizeEach
Showing 4 of 4
Improve instrument performance with the right selection of graphite cuvettesWhile ridged cuvettes are supplied as standard with Thermo Scientific GFS33/GFS35/GFS35Z Graphite Furnaces, additional application-specific cuvettes improve performance for more complex applications:
  • Extended Lifetime Cuvettes (ELCs) can be used longer and provide reproducible performance over several thousand firings
  • Ridged Cuvettes (in pyrolytically-coated and uncoated forms) help maintain difficult samples at the optimum temperature for efficient atomization
  • Omega Cuvettes with integral platforms provide the most interference-free conditions (often in combination with matrix modification) for samples with complex matrices