Search Thermo Fisher Scientific
Evaluate analytical performance at every stage of sample processing. Intended for use as an independent panel for analytical evaluation of BCR-ABL test methods. For research use only.
Description | AcroMetrix BCR-ABL Panel (RUO – Not for Clinical Use) |
Type | BCR-ABL Panel |
Packaging | One box of controls contains five panel members Panels A-E |
Quantity | 5 Box |
Target Gene | BCR-ABL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
956980TS | Each | AcroMetrix BCR-ABL Panel (RUO – Not for Clinical Use) | 1,000.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|