| 984368 | Full specifications | Each | Phosphate system reagents - R2 (3000 tests) | Request A Quote |
Type | Phosphate System Reagent | Sample Type | Water Sample, Soil Sample | Description | Phosphate system reagents - R2 (3000 tests) | Detects | Phosphate R2 | No. of Tests | 3000 | Test Range | 1.0 to 10.0 | Quantity | 4 x 20 mL | Interference Filter | 880 | Unit Size | Each |
|
| 984187 | Full specifications | Each | TON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests) | Request A Quote |
Type | TON Enzymatic Reduction System Reagent | Sample Type | Water Sample, Soil Sample | Description | TON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests) | Detects | TON Enz | No. of Tests | 275 | Test Range | 0 to 25 (for Gallery), 0 to 50 (for Aquakem) | Capacity (Metric) | 20 mL | Quantity | 20 mL | Interference Filter | 540/700 (for Gallery) 570 (for Aquakem) | Unit Size | Each |
|
| 984350 | Full specifications | Each | TON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests) | Request A Quote |
Type | TON-V Vanadium Reduction System Reagent | Sample Type | Water Sample, Soil Sample | Description | TON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests) | Detects | TON-V R1 | No. of Tests | 360 | Test Range | 0.4 to 50 | Quantity | 4 x 10 mL | Interference Filter | 540 | Unit Size | Each |
|
| 984351 | Full specifications | Each | TON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests) | Request A Quote |
Type | TON-V Vanadium Reduction System Reagent | Sample Type | Water Sample, Soil Sample | Description | TON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests) | Detects | TON-V R2 | No. of Tests | 500 | Test Range | 0.4 to 50 | Quantity | 4 x 10 mL | Interference Filter | 540 | Unit Size | Each |
|
| 984357 | Full specifications | Pack of,4 | Chromium (VI) system reagents (480 tests) | Request A Quote |
Type | Chromium System Reagent | Sample Type | Water Sample, Soil Sample | Description | Chromium (VI) system reagents (480 tests) | Detects | Chromium (VI) | No. of Tests | 480 | Test Range | 0.1 to 1.0 | Quantity | 4 x 20 mL | Interference Filter | 540/ 880 | Unit Size | Pack of,4 |
|
| 984358 | Full specifications | Pack of,8 | Magnesium system reagents (350 tests) | Request A Quote |
Type | Magnesium System Reagent | Sample Type | Water Sample, Soil Sample | Description | Magnesium system reagents (350 tests) | Detects | Magnesium | No. of Tests | 350 | Test Range | 0 to 400 | Quantity | 8 x 11 mL | Interference Filter | 510 | Unit Size | Pack of,8 |
|
| 984362 | Full specifications | Each | Ammonia system reagents - R1 (2000 tests) | Request A Quote |
Type | Ammonia System Reagent | Sample Type | Water Sample, Soil Sample | Description | Ammonia system reagents - R1 (2000 tests) | Detects | Ammonia R1 | No. of Tests | 2000 | Test Range | 1.0 to 10.0 (for Gallery) | Capacity (Metric) | 125 mL | Quantity | 125 mL | Interference Filter | 660 | Unit Size | Each |
|
| 984363 | Full specifications | Each | Ammonia system reagents - R2 (1300 tests) | Request A Quote |
Type | Ammonia System Reagent | Sample Type | Water Sample, Soil Sample | Description | Ammonia system reagents - R2 (1300 tests) | Detects | Ammonia R2 | No. of Tests | 1300 | Test Range | 1.0 to 10.0 (for Gallery) | Quantity | 4 x 20 mL | Interference Filter | 660 | Unit Size | Each |
|
| 984364 | Full specifications | Each | Chloride system reagents - R1 (500 tests) | Request A Quote |
Type | Chloride System Reagent | Sample Type | Water Sample, Soil Sample | Description | Chloride system reagents - R1 (500 tests) | Detects | Chloride R1 | No. of Tests | 500 | Test Range | 100 to 500 (for Gallery), 100 to 1000 (for Aquakem) | Quantity | 4 x 20 mL | Interference Filter | 480 | Unit Size | Each |
|
| 984365 | Full specifications | Each | Chloride system reagents - R1 (2500 tests) | Request A Quote |
Type | Chloride System Reagent | Sample Type | Water Sample, Soil Sample | Description | Chloride system reagents - R1 (2500 tests) | Detects | Chloride R1L | No. of Tests | 2500 | Test Range | 100 to 500 (for Gallery), 100 to 1000 (for Aquakem) | Analytical Method | Chloride | Quantity | 20 x 20 mL | Interference Filter | 480 | Unit Size | Each |
|