Thermo Scientific™
Gallery™ System Reagents for Environmental and Industrial Analysis
Catalog number: 984630
Have Questions?
Thermo Scientific™
Gallery™ System Reagents for Environmental and Industrial Analysis
Catalog number: 984630
Have Questions?
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery™ system reagents for environmental analysis.
Catalog Number
984630
Unit Size
Pack of,4
Description
Fluoride system reagents - R2 (4800 tests)
Price (USD)
Full specifications
TypeFluoride System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionFluoride system reagents - R2 (4800 tests)
DetectsFluoride R2
No. of Tests4800
Test Range0 to 5.0 (for Gallery), 0 to 2.0 (for Aquakem)
Quantity4 x 20 mL
Interference Filter600/ 880
Unit SizePack of,4
Catalog Number
Unit Size
Each
Description
TON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests)
Price (USD)
Full specifications
TypeTON Enzymatic Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests)
DetectsTON Enz
No. of Tests275
Test Range0 to 25 (for Gallery), 0 to 50 (for Aquakem)
Capacity (Metric)20 mL
Quantity20 mL
Interference Filter540/700 (for Gallery)
570 (for Aquakem)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests)
Price (USD)
Full specifications
TypeTON-V Vanadium Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests)
DetectsTON-V R1
No. of Tests360
Test Range0.4 to 50
Quantity4 x 10 mL
Interference Filter540
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)
Price (USD)
Full specifications
TypeTON-V Vanadium Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)
DetectsTON-V R2
No. of Tests500
Test Range0.4 to 50
Quantity4 x 10 mL
Interference Filter540
Unit SizeEach
Catalog Number
Unit Size
Pack of,4
Description
Chromium (VI) system reagents (480 tests)
Price (USD)
Full specifications
TypeChromium System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionChromium (VI) system reagents (480 tests)
DetectsChromium (VI)
No. of Tests480
Test Range0.1 to 1.0
Quantity4 x 20 mL
Interference Filter540/ 880
Unit SizePack of,4
Catalog Number
Unit Size
Pack of,8
Description
Magnesium system reagents (350 tests)
Price (USD)
Full specifications
TypeMagnesium System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionMagnesium system reagents (350 tests)
DetectsMagnesium
No. of Tests350
Test Range0 to 400
Quantity8 x 11 mL
Interference Filter510
Unit SizePack of,8
Catalog Number
Unit Size
Each
Description
Ammonia system reagents - R1 (2000 tests)
Price (USD)
Full specifications
TypeAmmonia System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionAmmonia system reagents - R1 (2000 tests)
DetectsAmmonia R1
No. of Tests2000
Test Range1.0 to 10.0 (for Gallery)
Capacity (Metric)125 mL
Quantity125 mL
Interference Filter660
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Ammonia system reagents - R2 (1300 tests)
Price (USD)
Full specifications
TypeAmmonia System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionAmmonia system reagents - R2 (1300 tests)
DetectsAmmonia R2
No. of Tests1300
Test Range1.0 to 10.0 (for Gallery)
Quantity4 x 20 mL
Interference Filter660
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Chloride system reagents - R1 (500 tests)
Price (USD)
Full specifications
TypeChloride System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionChloride system reagents - R1 (500 tests)
DetectsChloride R1
No. of Tests500
Test Range100 to 500 (for Gallery), 100 to 1000 (for Aquakem)
Quantity4 x 20 mL
Interference Filter480
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Chloride system reagents - R1 (2500 tests)
Price (USD)
Full specifications
TypeChloride System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionChloride system reagents - R1 (2500 tests)
DetectsChloride R1L
No. of Tests2500
Test Range100 to 500 (for Gallery), 100 to 1000 (for Aquakem)
Analytical MethodChloride
Quantity20 x 20 mL
Interference Filter480
Unit SizeEach
Showing 10 of 30Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
984630Full specifications
Pack of,4Fluoride system reagents - R2 (4800 tests)Request A Quote
TypeFluoride System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionFluoride system reagents - R2 (4800 tests)
DetectsFluoride R2
No. of Tests4800
Test Range0 to 5.0 (for Gallery), 0 to 2.0 (for Aquakem)
Quantity4 x 20 mL
Interference Filter600/ 880
Unit SizePack of,4
984187Full specifications
EachTON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests)Request A Quote
TypeTON Enzymatic Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by enzymatic reduction system reagents - R1 (275 tests)
DetectsTON Enz
No. of Tests275
Test Range0 to 25 (for Gallery), 0 to 50 (for Aquakem)
Capacity (Metric)20 mL
Quantity20 mL
Interference Filter540/700 (for Gallery)
570 (for Aquakem)
Unit SizeEach
984350Full specifications
EachTON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests)Request A Quote
TypeTON-V Vanadium Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by vanadium reduction system reagents - R1 (360 tests)
DetectsTON-V R1
No. of Tests360
Test Range0.4 to 50
Quantity4 x 10 mL
Interference Filter540
Unit SizeEach
984351Full specifications
EachTON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)Request A Quote
TypeTON-V Vanadium Reduction System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionTON (nitrate + nitrite) by vanadium reduction system reagents - R2 (500 tests)
DetectsTON-V R2
No. of Tests500
Test Range0.4 to 50
Quantity4 x 10 mL
Interference Filter540
Unit SizeEach
984357Full specifications
Pack of,4Chromium (VI) system reagents (480 tests)Request A Quote
TypeChromium System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionChromium (VI) system reagents (480 tests)
DetectsChromium (VI)
No. of Tests480
Test Range0.1 to 1.0
Quantity4 x 20 mL
Interference Filter540/ 880
Unit SizePack of,4
984358Full specifications
Pack of,8Magnesium system reagents (350 tests)Request A Quote
TypeMagnesium System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionMagnesium system reagents (350 tests)
DetectsMagnesium
No. of Tests350
Test Range0 to 400
Quantity8 x 11 mL
Interference Filter510
Unit SizePack of,8
984362Full specifications
EachAmmonia system reagents - R1 (2000 tests)Request A Quote
TypeAmmonia System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionAmmonia system reagents - R1 (2000 tests)
DetectsAmmonia R1
No. of Tests2000
Test Range1.0 to 10.0 (for Gallery)
Capacity (Metric)125 mL
Quantity125 mL
Interference Filter660
Unit SizeEach
984363Full specifications
EachAmmonia system reagents - R2 (1300 tests)Request A Quote
TypeAmmonia System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionAmmonia system reagents - R2 (1300 tests)
DetectsAmmonia R2
No. of Tests1300
Test Range1.0 to 10.0 (for Gallery)
Quantity4 x 20 mL
Interference Filter660
Unit SizeEach
984364Full specifications
EachChloride system reagents - R1 (500 tests)Request A Quote
TypeChloride System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionChloride system reagents - R1 (500 tests)
DetectsChloride R1
No. of Tests500
Test Range100 to 500 (for Gallery), 100 to 1000 (for Aquakem)
Quantity4 x 20 mL
Interference Filter480
Unit SizeEach
984365Full specifications
EachChloride system reagents - R1 (2500 tests)Request A Quote
TypeChloride System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionChloride system reagents - R1 (2500 tests)
DetectsChloride R1L
No. of Tests2500
Test Range100 to 500 (for Gallery), 100 to 1000 (for Aquakem)
Analytical MethodChloride
Quantity20 x 20 mL
Interference Filter480
Unit SizeEach
Showing 10 of 30Show more
Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water