Invitrogen™

AbC™ Total Antibody Compensation Bead Kit

Catalog number: A10513
Have Questions?
Invitrogen™

AbC™ Total Antibody Compensation Bead Kit

Catalog number: A10513
Have Questions?
Catalog Number
A10513
Unit Size
25 tests
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A1051325 tests
-
A10497100 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The AbC™ Total Antibody Compensation Bead Kit provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated mouse, rat, hamster, or rabbit antibodies with any laser in flow cytometry.

• Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
• Brightest signal—most sensitive antibody-capture beads available
• Maximum laser compatibility—for use with all lasers

Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Often, however, cellular-based compensation controls are not completely effective, as many antigens are not highly expressed, and dimly stained cells can lead to inaccurate compensation settings. The AbC™ Total Antibody Compensation Bead Kit was developed to address this issue.

View a selection guide for all flow cytometry compensation beads.

How it works
Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads). After mixing with a fluorophore-conjugated mouse, rat, hamster, or rabbit antibody, the two components provide a distinct high-signal positive control with an appropriate negative population that can then be used to set compensation properly regardless of the intensity of the cells in the actual experiment.

Broadest multispecies reactivity
The AbC™ Total Antibody Compensation Bead Kit is ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3, IgM), rat (IgG1, IgG2a, IgG2b, IgG2c, IgM), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.

Brightest signal
The AbC™ Total Compensation Bead Kit demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.

Maximum laser compatibility
All AbC™ compensation bead kits are ideal for use with any excitation source, including the 405 nm laser which has historically provided higher background signals due to auto-fluorescence with other kits on the market.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
Diameter (Metric)
6 μm
Excitation Wavelength Range
any lasers (UV to 633/635)
For Use With (Equipment)
Flow Cytometer
Format
Tube
No. of Tests
25 Tests
Shipping Condition
Room Temperature
Product Line
AbC™
Product Type
Compensation Beads

Contents & Storage

Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text