Gibco™

CD OptiCHO™ AGT™ Medium

Catalog number: A1122205
Have Questions?
Gibco™

CD OptiCHO™ AGT™ Medium

Catalog number: A1122205
Have Questions?
Catalog Number
A1122205
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A112220510 L
-
A11222041 L
-
A1122201100 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO’s legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advanced CHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.

When to Use CD OptiCHO™ Medium
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky,” i.e. hard to grow or adapt

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

Consider CD CHO medium when:
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

Consider CD DG44 medium when:
• You use parental DG44 or DXB11 CHO cells (dhfr-)

Liquid Media Formulations
Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.

Specifications

Culture Environment
CO2
Cell Line
CHO
Classification
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type
Suspension Cell Culture
Form
Powder (AGT)
Product Type
CD OptiCHO AGT Medium
Serum Level
Serum-free
Sterility
Non-sterile
With Additives
High Glucose, Sodium Pyruvate, Sodium Bicarbonate
Without Additives
No Glutamine, No Phenol Red
Endotoxin Level
Low
Format
Securitainer Capped Vial
Included Antibiotics
No antibiotics
Inorganic Salts
Magnesium, Sodium Phosphate
Product Line
AGT™, Gibco™
Purity or Quality Grade
Certified
Quantity
10 L
Reconstituted Volume
10 L
Remarks
Medium does not support wild type DG44 cells
Shipping Condition
Wet Ice

Contents & Storage

1 x 10 L, Storage 2-8°C Dark and Dry

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text