Informational:Interested in cGMP-banked Expi293F cells? Click here for product information.
Gibco™

Expi293™ Expression Medium

Catalog number: A1435103
Have Questions?
Gibco™

Expi293™ Expression Medium

Catalog number: A1435103
Have Questions?
Catalog Number
A1435103
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A143510310 L
-
A143510420 L
-
A14351011,000 mL
-
A14351026 x 1 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.

• Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
• Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
• Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
• Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10L in disposable bioreactors
• Does not contain phenol red

Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

Product use
Customers using Expi293 Expression Medium in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Cell Line
Expi293F™
Classification
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form
Liquid
Product Type
Expression Medium
With Additives
GlutaMAX
Without Additives
No Phenol Red
Product Line
Expi293™
Quantity
10 L

Contents & Storage

One 10L Universal Bag
Store in the dark at 2-8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text