Conceptual art depiciting CRISPR screening demonstrated by endless rows of plates and a human hand selecting specific wells
Invitrogen LentiArray CRISPR gRNA are lentiviruses designed for efficient gene editing in your CRISPR-Cas9 experiments, including high-throughput screening and editing of hard-to-transfect cell lines. Select from our suite of Invitrogen LentiArray CRISPR gRNA using the tool below.
LentiArray CRISPR gRNA selection tool
Product Type
Species
Can't find what you are looking for? Contact us for more information about CRISPR products and services.
CRISPR Controls
High quality controls play an integral role in the successful development and performance of high throughput screens. Be sure to obtain controls to help optimize delivery conditions, maximize editing efficiency, and establish hit selection criteria.
Explore CRISPR Controls