Thermo Scientific™
Thermo Scientific™ S1 Pipet Filler and Nunc™ Serological Pipettes
Catalog number: A33236
This combination of Thermo Scientific™ S1 Pipet Filler and Nunc™ Serological Pipettes offers a convenient start to pipetting productivity in cell culture and large volume pipetting applications.
The S1 Pipet Filler is a motorized filler with these features:
- LCD display with speed and battery indicators
- Separate speed control for aspiration and dispensing
- Lithium-ion battery
- Industry-leading 2-year warranty with web registration
- A choice of five colors (white, blue, green, clear, red)
Nunc Serological Pipettes are disposable and individually wrapped pipettes with these features:
- 100% virgin polystyrene for maximum clarity
- Certified non-pyrogenic
- Bold, clear graduations with easy-to-read scale markings
- Non-dripping tip for pipette precision
- Color-coded packaging for ease in sorting and selecting by size
- Individually wrapped; paper/plastic or plastic film; peel or pop through for easy opening
- Bulk packed for frequent use; reduces packaging waste
- A choice of two sizes (10 mL and 25 mL)