Precision ID IonCode™ Barcode Adapters 1–96 Kit in 96-Well PCR Plate
Precision ID IonCode™ Barcode Adapters 1–96 Kit in 96-Well PCR Plate
Applied Biosystems™
Precision ID IonCode™ Barcode Adapters 1–96 Kit in 96-Well PCR Plate
The Precision ID IonCode Barcode Adapters 1–96 Kit in 96-Well PCR Plate provides a set of 96 unique barcode adaptersRead more
Have Questions?
Catalog NumberQuantity
A33586960 Reactions
Catalog number A33586
Price (HKD)
29,566.00
Each
Add to cart
Quantity:
960 Reactions
Price (HKD)
29,566.00
Each
Add to cart
Ask our AI about this Product
The Precision ID IonCode Barcode Adapters 1–96 Kit in 96-Well PCR Plate provides a set of 96 unique barcode adapters specifically designed for optimal performance with Precision ID library kits and panels and Ion S5 or Ion S5 XL Systems. When used in combination with the Precision ID Library Kit, the Precision ID IonCode Barcode Adapters 1–96 Kit in 96-Well PCR Plate allows users to pool amplicon libraries prepared manually or with automation and conduct multiplexed sequencing analysis, thereby increasing lab efficiency and reducing sequencing costs per sample.
Key product features:
• Robust barcodes ligate efficiently and uniformly to generate balanced representation of libraries
• Unbiased performance helps ensure identical genotypes independent of barcode used
• Robust error correction built in by design for added confidence in sample identification
Increase sample multiplexing without impacting data quality
Multiplexing with barcoded libraries leads to more cost effective sequencing. Each barcode has been individually evaluated to help ensure no bias of sequence context representation. The barcodes were also screened for equivalent efficiency to enable balanced read depth between libraries.
Specifications
Format96-well Plate
Product TypeBarcode Adapters 1-96 Kit
Quantity960 Reactions
Sequencing TypeGenome & DNA Sequencing
Shipping ConditionDry Ice
Product LineIonCode™
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Have questions about this product? Ask our AI assisted search.
Where can I find the formulation of MEM Non-Essential Amino Acids Solution (100X) (Cat. No. 11140050)?
What is Gibco MEM Non-Essential Amino Acids?
What is the solvent used for MEM Non-Essential Amino Acids Solution (100X)?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Los clientes que vieron este artículo también vieron
Frequently asked questions (FAQs)
The Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate contains 20 µL/well of each barcode.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
The Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate provides 10 reactions per barcode.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
The Precision ID IonCode Barcode Adapters 1-96 Kit in 96-Well PCR Plate provides enough reagents for 960 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Los productos recomendados
Share catalog number, name or link
1x1 image pixel for data collection