Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | TMT10plex Isobaric Label Reagent Set plus TMT11-131C Label Reagent, 1 x 5 mg (per tag) |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag (TMT) Label |
Shipping Condition | Dry Ice |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide Labeling |
Quantity | 1 x 5 mg (per tag) |
Unit Size | 1 x 5 mg (per tag) |
Description | TMT11-131C Label Reagent, 1 x 5 mg |
Product Type | Isobaric Label Reagent |
Label or Dye | Tandem Mass Tag (TMT) Label |
Shipping Condition | Dry Ice |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide labeling and quantitation |
Quantity | 1 x 5 mg |
Unit Size | 1 x 5 mg |
Description | TMT11-131C Label Reagent, 3 x 0.8 mg |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent |
Chemical Reactivity | Amine |
Label or Dye | Tandem Mass Tag (TMT) Label |
Shipping Condition | Dry Ice |
Starting Material | Peptides |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide Labeling and quantitation |
Quantity | 3 x 0.8 mg |
Form | Solid |
Unit Size | 3 x 0.8 mg |
Description | TMT10plex Isobaric Label Reagent Set plus TMT11-131C Label Reagent, 3 x 0.8 mg (per tag) |
For Use With (Equipment) | Mass Spectrometer |
Product Line | TMT10plex™ |
Label or Dye | Mass Isotopes of Amino Acids |
Shipping Condition | Dry Ice |
Starting Material | Peptides |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide Labeling |
Quantity | 3 x 0.8 mg (per tag) |
Unit Size | 3 x 0.8 mg (per tag) |
Description | TMT11plex Label Reagents in stabilized solution, 96-well plate, 8 x 40 μg (per tag) |
Formulation | Liquid in stabilized DMSO solution |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT10plex™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein PEPTIDE |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 88 Reactions |
Form | Liquid |
Unit Size | 8 x 40 μg (per tag) |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A34808 | 1 x 5 mg (per tag) | TMT10plex Isobaric Label Reagent Set plus TMT11-131C Label Reagent, 1 x 5 mg (per tag) | 5,732.65 Online price 6,300.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A34807 | 1 x 5 mg | TMT11-131C Label Reagent, 1 x 5 mg | 488.65 Online price 536.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A37724 | 3 x 0.8 mg | TMT11-131C Label Reagent, 3 x 0.8 mg | 357.65 Online price 367.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A37725 | 3 x 0.8 mg (per tag) | TMT10plex Isobaric Label Reagent Set plus TMT11-131C Label Reagent, 3 x 0.8 mg (per tag) | 3,251.65 Online price 3,720.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A58333 | 8 x 40 μg (per tag) | TMT11plex Label Reagents in stabilized solution, 96-well plate, 8 x 40 μg (per tag) | 640.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The advantages of TMT label reagents include increased multiplexed relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents plus TMT11-131C is ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or to improve statistics on biological replicates.
Features of this TMT11-131C Label Reagent include:
• Multiplex—concurrent MS analysis of up to 11 samples derived from cells, tissues, or biological fluids
• Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
• Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
• Compatible—optimized for use with high resolution Thermo Scientific Orbirtrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3
TMT11-131C is available as a standalone tag (Cat. No. A34807, A37724), as part of this TMT10plex Isobaric Label Reagent Set plus TMT11-131C, or as part of the complete 11plex set of reagents in a 96-well plate format. When combined with the industry-leading, high resolution Orbitrap instruments and software, TMT reagents provide integrated total solutions for quantitative protein expression analysis.