Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Annexin V, Alexa Fluor 555 conjugate |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 555 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 532, 561 |
Excitation/Emission | 555/565 |
No. of Reactions | 100 |
Color | Orange |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 555 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, Alexa Fluor 488 conjugate (replacement for Cat. No. PHN1010 and PHN1008) |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 488 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 488 |
Excitation/Emission | 495/519 |
No. of Reactions | 100 |
Color | Green |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 488 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, FITC conjugate (replaces AnnexinV01, AnnexinV013, PHN1010, PHN1008) |
Kit Contents | Contains 1 vial of annexin V, FITC conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 488 |
Excitation/Emission | 494/518 |
No. of Reactions | 100 |
Color | Green |
Shipping Condition | Wet Ice |
Conjugate | FITC |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, Alexa Fluor 568 conjugate |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 568 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 532, 561 |
Excitation/Emission | 578/603 |
No. of Reactions | 100 |
Color | Orange-Red |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 568 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, Alexa Fluor 594 conjugate |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 594 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 532 |
Excitation/Emission | 590/617 |
No. of Reactions | 100 |
Color | Red |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 594 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, biotin-X conjugate |
Kit Contents | Contains 1 vial of annexin V, biotin-x conjugate. |
Product Type | Annexin V conjugate |
No. of Reactions | 100 |
Shipping Condition | Wet Ice |
Conjugate | Biotin-X |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, Alexa Fluor 350 conjugate |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 350 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | UV |
Excitation/Emission | 346/442 |
No. of Reactions | 100 |
Color | Blue |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 350 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, Alexa Fluor 647 conjugate |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 647 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 633-637 |
Excitation/Emission | 650/665 |
No. of Reactions | 100 |
Color | Far-Red |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 647 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, Alexa Fluor 680 conjugate |
Kit Contents | Contains 1 vial of annexin V, Alexa Fluor 680 conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 633-637 |
Excitation/Emission | 679/702 |
No. of Reactions | 100 |
Color | Near IR |
Shipping Condition | Wet Ice |
Conjugate | Alexa Fluor 680 |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 500 µL |
Description | Annexin V, APC conjugate (APC Annexin V) (replaces Annexin product- AnnexinV05) |
Kit Contents | Contains 1 vial of annexin V, APC conjugate. |
Product Type | Annexin V conjugate |
Flow Cytometer Laser Lines | 633-637 |
Excitation/Emission | 650/660 |
No. of Reactions | 50 |
Color | Red |
Shipping Condition | Wet Ice |
Conjugate | APC (Allophycocyanin) |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 250 µL |
Catalog Number | Specifications | Unit Size | Conjugate | Excitation/Emission | Flow Cytometer Laser Lines | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A35108 | 500 µL | Alexa Fluor 555 | 555/565 | 532, 561 | 388.65 Online price 437.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A13201 | 500 µL | Alexa Fluor 488 | 495/519 | 488 | 356.65 Online price 395.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A13199 | 500 µL | FITC | 494/518 | 488 | 358.65 Online price 395.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A13202 | 500 µL | Alexa Fluor 568 | 578/603 | 532, 561 | 397.65 Online price 437.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A13203 | 500 µL | Alexa Fluor 594 | 590/617 | 532 | 388.65 Online price 437.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A13204 | 500 µL | Biotin-X | 388.65 Online price 436.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A23202 | 500 µL | Alexa Fluor 350 | 346/442 | UV | 395.65 Online price 429.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A23204 | 500 µL | Alexa Fluor 647 | 650/665 | 633-637 | 391.65 Online price 437.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A35109 | 500 µL | Alexa Fluor 680 | 679/702 | 633-637 | 395.65 Online price 428.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A35110 | 250 µL | APC (Allophycocyanin) | 650/660 | 633-637 | 458.65 Online price 508.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|