Ion Torrent™

Ion AmpliSeq™ Transcriptome Mouse Gene Expression Kit

Catalog number: A36553
Have Questions?
Ion Torrent™

Ion AmpliSeq™ Transcriptome Mouse Gene Expression Kit

Catalog number: A36553
Have Questions?
Catalog Number
A36553
Unit Size
24 reactions
Price (USD)
Quantity
Catalog Number
Unit Size
96 reactions
Price (USD)
Quantity
Catalog Number
Unit Size
384 reactions
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A3655324 reactions
A3655496 reactions
A36555384 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion AmpliSeq Transcriptome Mouse Gene Expression Assay allows simultaneous gene expression measurement of over 20,000 mouse RefSeq genes in a single assay. Providing the ability to work directly with low input levels of total RNA, the Ion AmpliSeq Transcriptome Mouse Gene Expression Assay is ideal for those interested in obtaining comprehensive and highly sensitive gene expression measurements from a wide range of RNA sources. This particular kit enables manual library preparation with throughput flexibility. For automated production, see the Ion AmpliSeq Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit.

The Ion AmpliSeq Transcriptome Mouse Gene Expression Assay offers:
• Flexible, straightforward library preparation—start with as little as 10 ng of total RNA from tissues, cells, or other sources
• Fast gene expression profiling with simple workflow—go from RNA to data in <2 days with ∼45 minutes of total hands-on time with the automated workflow of the Ion GeneStudio S5 and Ion Chef systems
• A complementary software plug-in for automated sequencing analysis that provides gene quantitation and reporting as well as quality control metrics
• Compatibility with Ion Xpress Barcode adapters, allowing for multiplexed sequencing

Leveraging the speed and simplicity of Ion AmpliSeq technology, this ultrahigh-multiplex PCR enrichment approach enables rapid gene expression profiling with minimal hands-on time. This research method provides a faster, simpler, and lower-cost sequencing method that is highly correlated to whole-transcriptome RNA sequencing.

Straightforward library preparation and multiplex sequencing with manual kit
Starting with as little as 10 ng of total RNA, libraries can be prepared in approximately seven hours with only one hour of hands-on time. Total RNA is first processed with the SuperScript VILO cDNA Synthesis Kit (not included), and the resulting cDNA is amplified using Ion AmpliSeq technology to accurately maintain expression levels of all genes.

The resulting libraries are ready for downstream template preparation using the Ion OneTouch 2 or Ion Chef System and sequencing on the Ion GeneStudio S5 or Ion Proton System. Libraries can be barcoded for multiplexing, providing flexibility, reduced cost, and increased throughput, with the recommended 16 libraries per Ion 550 Chip and 8 libraries per Ion 540 or Ion PI chip. Other multiplexing options are also possible, depending on the experimental design and desired level of sensitivity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Kit Type
Transcriptome Mouse Gene Expression Kit
Libraries
Targeted Sequencing Library
Product Line
Ion AmpliSeq™
Shipping Condition
Dry Ice
For Use With (Application)
Gene Expression, miRNA analysis
No. of Reactions
24 Reactions
Starting Material
RNA

Contents & Storage

Store in freezer at -5 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text