Informational:Detect rhabdovirus contamination in Sf9 cultures. Learn more »
Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

ExpiSf™ CD Medium

Catalog number: A3767801
Have Questions?
Gibco™

ExpiSf™ CD Medium

Catalog number: A3767801
Have Questions?
Catalog Number
A3767801
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3767801500 mL
-
A37678021000 mL
-
A37678036 x 1000 mL
-
A376780410 L
-
A376780520 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
ExpiSf CD Medium is a GMP-manufactured, chemically defined, yeastolate-free, serum-free, protein-free, animal origin-free medium specifically designed for high-density growth, transfection, and baculovirus infection of suspension-adapted Sf9 (Spodoptera frugiperda) insect cells.

ExpiSf CD Medium:
• Delivers high reproducibility and lot-to-lot reliability
• Enables high-yield protein expression that is three times higher than traditional insect expression platforms
• Supports growth of suspension Sf9 cultures to densities of approximately 2 x 107 cells/mL
• Supports high-density baculovirus infection of suspension Sf9 cells
• Supports growth, transfection, and baculovirus infection of Sf9 cells in culture formats from 24-deep well plates to >2 L in shaker flasks
• Is a ready-to-use formulation, with no supplementation required
• Powder format available to facilitate large-scale applications

ExpiSf CD Medium is a core component of the ExpiSf Expression System. For highest level protein expression, the medium is used in combination with a matched protein expression enhancer, ExpiSf Enhancer, which is supplied as part of the ExpiSf Protein Production Kit. Sf9 and Sf21 cells adapted to other commercially-available serum-free media may be adapted for growth in ExpiSf CD Medium. ExpiSf CD Medium is not recommended for adherent cell cultures.

cGMP manufacturing
ExpiSf CD Medium is manufactured using an ISO 13485-certified process according to 21 CFR 820 medical device current good manufacturing practices (cGMP).

Dry media formulations
For added flexibility, ExpiSf CD Medium can be made in AGT dry format at large scale. For custom media requests, please contact Gibco Custom Media Services.

Specifications

Quantity
500 mL
Classification
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Compatible Cells
Insect Cells
Format
Liquid
Shipping Condition
Room Temperature
Product Line
ExpiSf™

Contents & Storage

1 x 500 mL bottle, store in the dark at 2–8°C

Shelf Life: 12 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text