Gibco™

ExpiSf™ Protein Production Kit

Catalog number: A3767806
Have Questions?
Gibco™

ExpiSf™ Protein Production Kit

Catalog number: A3767806
Have Questions?
Catalog Number
A3767806
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A37678061 L
-
A37678096 x 1 L
-
A376780710 L
-
A3767808
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The ExpiSf Protein Production Kit is designed to significantly boost protein expression from high-density suspension cultures of Sf9 insect cells. The kit includes the chemically defined, animal origin-free ExpiSf Enhancer and ExpiSf CD Medium, which together power the highest possible level of protein expression from baculovirus-infected ExpiSf9 Cells.

The ExpiSf Protein Production Kit:
• Delivers greater than 80% more protein yield in high-density suspension Sf9 cell culture than ExpiSf CD Medium alone
• Includes ExpiSf Enhancer, which is specifically designed to work in concert with ExpiSf CD Medium to significantly boost protein expression from baculovirus-infected insect cell cultures
• Demonstrates direct scalability of baculovirus infection from culture volumes of 4 mL in deep-well plates to >3 L in shake flasks

The ExpiSf Protein Production Kit is specifically designed to work with ExpiSf9 Cells in the ExpiSf Expression System. The kit supplies sufficient ExpiSf Enhancer and ExpiSf CD Medium to infect 1 L, 10 L, or 50 L of culture. ExpiSf CD Medium is also available for purchase separately.

Specifications

Type
Media and reagents, ExpiSf CD Meidum and ExpiSf Enhancer
Quantity
1 L (ExpiSf CD Medium), 4 x 1 mL (ExpiSf Enhancer)
Classification
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Compatible Cells
Insect Cells
Format
Liquid
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
Expression Mechanism
Cell-Based Expression
Expression System
Baculovirus
Product Line
ExpiSf™

Contents & Storage

1000 mL bottle ExpiSf CD Medium, store in the dark at 2°C to 8°C; 4 x 1 mL ExpiSf Enhancer, store (at 2°C to 8°C; Shelf Life: 12 months from date of manufacture)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text