Thermo Scientific™

SGS™ All Species ID Food DNA Analyser Kits

Catalog number: A38455
Have Questions?
Thermo Scientific™

SGS™ All Species ID Food DNA Analyser Kits

Catalog number: A38455
Have Questions?

Identify the species present in even the most challenging sample types with the Thermo Scientific™ Next Generation Sequencing (NGS) Food Authenticity Workflow incorporating SGS™ All Species ID Food DNA Analyser Kits.

Protect your business from food fraud by verifying the meat, plant or fish species in your food samples for improved traceability and food safety.

As part of a complete, simple Next Generation Sequencing workflow, these kits provide a list of meat, plant or fish species present in all types of food and feed products including complex and highly processed sample types.

 
Catalog Number
A38455
Unit Size
Each
Description
SGS All Species ID FISH DNA Analyser Kit II
Barcode
25-48
Price (USD)
Full specifications
Barcode25-48
DescriptionSGS All Species ID FISH DNA Analyser Kit II
FormatKit
Sample TypeDNA
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SGS All Species ID MEAT DNA Analyser Kit I 
Barcode
1-24
Price (USD)
Full specifications
Barcode1-24
DescriptionSGS All Species ID MEAT DNA Analyser Kit I 
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SGS All Species ID MEAT DNA Analyser Kit II
Barcode
25-48
Price (USD)
Full specifications
Barcode25-48
DescriptionSGS All Species ID MEAT DNA Analyser Kit II
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SGS All Species ID FISH DNA Analyser Kit I
Barcode
1-24
Price (USD)
Full specifications
Barcode1-24
DescriptionSGS All Species ID FISH DNA Analyser Kit I
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SGS All Species ID PLANT DNA Analyser Kit I
Barcode
1-24
Price (USD)
Full specifications
Barcode1-24
DescriptionSGS All Species ID PLANT DNA Analyser Kit I
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SGS All Species ID PLANT DNA Analyser Kit II
Barcode
25-48
Price (USD)
Full specifications
Barcode25-48
DescriptionSGS All Species ID PLANT DNA Analyser Kit II
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
Catalog Number
Unit Size
Each
Description
SGS All Species ID Software
Barcode
Price (USD)
Full specifications
DescriptionSGS All Species ID Software
FormatKit
For Use With (Application)Sequencing
Product TypeAll Species ID Food DNA Analyzer Kit
Quantity2 Licenses
SpeciesAll Species
Unit SizeEach
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeDescriptionBarcodePrice (USD)
A38455Full specifications
EachSGS All Species ID FISH DNA Analyser Kit II25-48Request A Quote
Barcode25-48
DescriptionSGS All Species ID FISH DNA Analyser Kit II
FormatKit
Sample TypeDNA
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
A38452Full specifications
EachSGS All Species ID MEAT DNA Analyser Kit I 1-24Request A Quote
Barcode1-24
DescriptionSGS All Species ID MEAT DNA Analyser Kit I 
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
A38453Full specifications
EachSGS All Species ID MEAT DNA Analyser Kit II25-48Request A Quote
Barcode25-48
DescriptionSGS All Species ID MEAT DNA Analyser Kit II
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
A38454Full specifications
EachSGS All Species ID FISH DNA Analyser Kit I1-24Request A Quote
Barcode1-24
DescriptionSGS All Species ID FISH DNA Analyser Kit I
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
A38456Full specifications
EachSGS All Species ID PLANT DNA Analyser Kit I1-24Request A Quote
Barcode1-24
DescriptionSGS All Species ID PLANT DNA Analyser Kit I
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
A38457Full specifications
EachSGS All Species ID PLANT DNA Analyser Kit II25-48Request A Quote
Barcode25-48
DescriptionSGS All Species ID PLANT DNA Analyser Kit II
FormatKit
For Use With (Application)Sequencing
No. of Reactions96 Tests
Packaging Type1 box
Product TypeAll Species ID Food DNA Analyzer Kit
Shelf Life52 weeks
SpeciesAll Species
Unit SizeEach
A38458Full specifications
EachSGS All Species ID SoftwareRequest A Quote
DescriptionSGS All Species ID Software
FormatKit
For Use With (Application)Sequencing
Product TypeAll Species ID Food DNA Analyzer Kit
Quantity2 Licenses
SpeciesAll Species
Unit SizeEach
Showing 7 of 7

Identification of the species present in food and feed samples is a critical step for proof of authenticity, verification of origin, traceability of raw materials and quality control of handling and cleaning processes in food production lines.

The Thermo Scientific NGS Food Authenticity Workflow utilizes the Ion Torrent™ Next Generation Sequencing technology to enable an untargeted screening approach making it possible to identify the species contained in a food sample by comparison with a DNA database of meat, plant or fish species.

The ready-to-use SGS All Species ID Food DNA Analyser Kits allow the unique labelling and amplification of meat, fish or plant DNA by PCR, thereby preparing the sample for Next Generation Sequencing, and automated data analysis by the SGS All Species ID Software. Test results provide a list of the meat, plant or fish species present in the sample.

  • Simple: ready-to-use kits include all necessary reagents for DNA labeling and amplification, and provide results with any sample type, including complex and processed samples.
  • Rapid: allows preparation of NGS library from extracted DNA within 4 hours.
  • Flexible: utilizes a unique labeling system in which the DNA of each food sample is individually identified with an exclusive barcoded DNA sequence, enabling you to analyze multiple samples together for meat, fish or plant species.  
  • Supported: our experienced technical team is ready to assist you.

Each SGS All Species ID Food DNA Analyser Kit is available in two different formats with 24 different barcodes each, enabling you to label up to 48 samples at once for multiple simultaneous sample analysis.

For Laboratory Use

Contents & Storage

≤-18°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text