Collibri™ PCR-free ES DNA Library Prep Kits for Illumina Systems
Catalog number: A38545024W
Have Questions?
Invitrogen™
Collibri™ PCR-free ES DNA Library Prep Kits for Illumina Systems
Catalog number: A38545024W
Have Questions?
The Collibri PCR-free ES DNA Library Preparation Kit is designed for fast and convenient library preparation from inputs as low as 100 ng intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about two hours for the PCR-free workflow. If needed, Collibri Library Amplification Master Mix with primers can be bought separately. Or see the Collibri ES DNA Library Preparation Kit, designed for fast and convenient library preparation with PCR from a wide range of inputs (1–500 ng).
Whole Genome Sequencing, Resequencing, De Novo Assembly
Includes
A39096096; A39123196; A38584096
Libraries
Fragment Library
No. of Reactions
96 Preps
Product Line
Collibri™
Product Type
PCR-free ES DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
Quantity
96 preps
Shipping Condition
Cool Pack & Dry Ice
Species
All
Workflow Step
Library Generation
Starting Material Amount
100-500 ng
Unit Size
96 reactions
Showing 9 of 9
The Collibri PCR-free ES DNA Library Preparation Kit is designed for fast and convenient library preparation from inputs as low as 100 ng intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about two hours for the PCR-free workflow. If needed, Collibri Library Amplification Master Mix with primers can be bought separately. Or see the Collibri ES DNA Library Preparation Kit, designed for fast and convenient library preparation with PCR from a wide range of inputs (1–500 ng).
The Collibri PCR-free ES DNA Library Preparation Kit: • Includes all necessary components in a single kit* for quick use • Is empowered on miniaturization liquid handlers to cut on costs • Provides in-process visual feedback for easier handling •Is suitable for conventional automation to increase throughput
* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters
The Collibri PCR-free ES DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. Moreover, the enzymatic shearing (ES) module of the kit generates a consistently high yield of library, which is enough for library quantification and sequencing and multiple runs on Illumina instruments.
Applications The Collibri PCR-free ES DNA Library Preparation Kit is recommended for: • Whole genome sequencing • Resequencing • De novo assembly
For Research Use Only. Not for use in diagnostic procedures.
Contents & Storage
• Collibri PCR-free ES DNA Library Prep Kit, store at -20°C • Collibri DNA Library Cleanup Kit, store at 4°C • Collibri DNA CD Indexes, store at -20°C