Informational:資訊性:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Collibri™ PS DNA Library Prep Kits for Illumina Systems

產品號碼: A38614196
Invitrogen™

Collibri™ PS DNA Library Prep Kits for Illumina Systems

產品號碼: A38614196
The Collibri PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.
 
產品號碼
A38614196
Unit Size
96 preps
Product Type
PS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
No. of Reactions
96 Preps
供應狀況
-
完整規格
數量
-
DescriptionThe product contains modules A39099096, A39123196, A38584096
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions96 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
Quantity96 preps
RemarksThe product contains modules A39099096, A39123196, A38584096
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size96 preps
產品號碼
Unit Size
24 preps
Product Type
PS DNA Library Prep Core Kit
No. of Reactions
24 Preps
價格 (TWD)
價格 31,900.00
線上優惠 22,330.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39122024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Core Kit
Quantity24 preps
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit Size24 preps
產品號碼
Unit Size
96 preps
Product Type
PS DNA Library Prep Core Kit
No. of Reactions
96 Preps
價格 (TWD)
價格 122,600.00
線上優惠 85,820.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39099096, A38584096
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions96 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Core Kit
Quantity96 preps
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit Size96 preps
產品號碼
Unit Size
24 preps
Product Type
PS DNA Library Prep Kit with CD Indexes
No. of Reactions
24 Preps
價格 (TWD)
價格 46,600.00
線上優惠 32,620.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39122024, A38586024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with CD Indexes
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
產品號碼
Unit Size
96 preps
Product Type
PS DNA Library Prep Kit with CD Indexes
No. of Reactions
96 Preps
價格 (TWD)
價格 164,700.00
線上優惠 131,760.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39099096, A39123096, A38584096
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions96 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with CD Indexes
Quantity96 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size96 preps
產品號碼
Unit Size
24 preps
Product Type
PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
No. of Reactions
24 Preps
價格 (TWD)
價格 50,400.00
線上優惠 35,280.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39122024, A38587024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
產品號碼
Unit Size
24 preps
Product Type
PS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
No. of Reactions
24 Preps
價格 (TWD)
價格 49,500.00
線上優惠 34,650.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39122024, A43348024 , A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
產品號碼
Unit Size
24 preps
Product Type
PS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
No. of Reactions
24 Preps
價格 (TWD)
價格 49,500.00
線上優惠 34,650.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39122024, A43350024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
Quantity24 preps
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit Size24 preps
產品號碼
Unit Size
24 preps
Product Type
PS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
No. of Reactions
24 Preps
價格 (TWD)
價格 49,500.00
線上優惠 34,650.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
完整規格
數量
DescriptionThe product contains modules A39122024, A43351024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
顯示 9 項,共 9 項
產品號碼規格Unit SizeProduct TypeNo. of Reactions價格 (TWD)供應狀況數量
A38614196完整規格
96 prepsPS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)96 Preps與我們聯繫 ›--
DescriptionThe product contains modules A39099096, A39123196, A38584096
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions96 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
Quantity96 preps
RemarksThe product contains modules A39099096, A39123196, A38584096
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size96 preps
A38612024W完整規格
24 prepsPS DNA Library Prep Core Kit24 Preps
價格 31,900.00
線上優惠 22,330.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39122024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Core Kit
Quantity24 preps
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit Size24 preps
A38614096W完整規格
96 prepsPS DNA Library Prep Core Kit96 Preps
價格 122,600.00
線上優惠 85,820.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39099096, A38584096
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions96 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Core Kit
Quantity96 preps
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit Size96 preps
A38612024完整規格
24 prepsPS DNA Library Prep Kit with CD Indexes24 Preps
價格 46,600.00
線上優惠 32,620.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39122024, A38586024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with CD Indexes
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
A38614096完整規格
96 prepsPS DNA Library Prep Kit with CD Indexes96 Preps
價格 164,700.00
線上優惠 131,760.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39099096, A39123096, A38584096
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions96 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with CD Indexes
Quantity96 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size96 preps
A38613024完整規格
24 prepsPS DNA Library Prep Kit with UD Indexes (Set A, 1-24)24 Preps
價格 50,400.00
線上優惠 35,280.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39122024, A38587024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
A43611024完整規格
24 prepsPS DNA Library Prep Kit with UD Indexes (Set B, 25-48)24 Preps
價格 49,500.00
線上優惠 34,650.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39122024, A43348024 , A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
A43612024完整規格
24 prepsPS DNA Library Prep Kit with UD Indexes (Set C, 49-72)24 Preps
價格 49,500.00
線上優惠 34,650.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39122024, A43350024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
Quantity24 preps
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit Size24 preps
A43613024完整規格
24 prepsPS DNA Library Prep Kit with UD Indexes (Set D, 73-96)24 Preps
價格 49,500.00
線上優惠 34,650.00
(01-Oct-2024 結束)
您的價格
-
DescriptionThe product contains modules A39122024, A43351024, A43190024
FormatKit
Sample TypeDNA
Sequencing TypeGenome and DNA Sequencing
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 & 2000, NovaSeq 6000
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri™
Product TypePS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
Quantity24 preps
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
Unit Size24 preps
顯示 9 項,共 9 項

The Collibri PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PS DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters.

The Collibri PS DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. The kit generates a consistently high yield of library from any amount of input DNA (including the PCR-free version), which is enough for library quantification   and sequencing on Illumina instruments.

Applications
The Collibri PS DNA Library Preparation Kit is recommended for:
Whole genome sequencing
• Resequencing
• De novo assembly

For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

內容物與存放

• Collibri PS DNA Library Prep Kit, store at -20°C
• Collibri DNA Library Cleanup Kit, store at 4°C
• Collibri DNA UD Indexes, store at -20°C

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text