Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

EquiPhi29™ DNA Polymerase

Catalog number: A39390
Have Questions?
Thermo Scientific™

EquiPhi29™ DNA Polymerase

Catalog number: A39390
Have Questions?
Catalog Number
A39390
Unit Size
250 U
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 U
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5000 U
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A39390250 U
-
A393911000 U
-
A393925000 U
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution. This enzyme is significantly improved over phi29 DNA polymerase in protein thermostability, reaction speed, product yield, and amplification bias, while retaining all the benefits of the wild-type enzyme.

EquiPhi29 DNA Polymerase possesses strong strand displacement activity and 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Features of EquiPhi29 DNA Polymerase
• Highest processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
• Low amplification bias
• Extremely high yields of amplified DNA, even from small amounts of template
• Highly accurate DNA synthesis in a short time 

Applications
EquiPhi29 DNA Polymerase may be used in a wide variety of applications including:
• Unbiased whole genome amplification (WGA)
• Rolling circle amplification (RCA)
• Protein-primed DNA amplification
• Cell-free cloning of lethal DNA
In situ genotyping with padlock probes
• RNA-primed DNA amplification

Notes
1. The addition of pyrophosphatase to the reaction mixture with EquiPhi29 DNA Polymerase may further enhance DNA synthesis
2. Because of the enzyme's 3'→5' proofreading exonuclease, 3’-modified primers are highly recommended.
3. EquiPhi29 DNA Polymerase can amplify DNA in a wide range of temperatures (30–45°C), with an optimal reaction temperature of 42°C.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Stand-alone enzyme
For Use With (Application)
MDA-WGA, RCA
Polymerase
EquiPhi29 DNA Polymerase
Quantity
250 U
Optimal Reaction Temperature
42°C
Shipping Condition
Dry Ice
Concentration
10 U/μL
Reaction Time
2 hours

Contents & Storage

• EquiPhi29 DNA Polymerase (25 μL at 10 U/μL)
• 10X EquiPhi29 DNA Polymerase Reaction Buffer (0.25 mL)
• 110 mM DTT (0.25 mL)

Store at –15 to –25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text