Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Ponceau S Staining Solution, 3 x 1000 mL |
Detection Method | Colorimetric |
Formulation | 0.1% (w/v) in 5% acetic acid |
For Use With (Application) | Western Blotting |
Quantity | 3 x 1000 mL |
Unit Size | 3 x 1 L |
Description | Ponceau S Staining Solution, 500 mL |
Detection Method | Colorimetric |
Form | Liquid |
Formulation | 0.1% (w/v) in 5% acetic acid |
For Use With (Application) | Western Blotting |
Quantity | 500 mL |
Unit Size | 500 mL |
Description | Ponceau S Staining Solution, 1000 mL |
Detection Method | Colorimetric |
Form | Liquid |
Formulation | 0.1% (w/v) in 5% acetic acid |
For Use With (Application) | Western Blotting |
Quantity | 1000 mL |
Unit Size | 1000 mL |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A40000278X3 | 3 x 1 L | Ponceau S Staining Solution, 3 x 1000 mL | 241.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A40000279 | 500 mL | Ponceau S Staining Solution, 500 mL | 71.65 Online price 73.25 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A40000278 | 1000 mL | Ponceau S Staining Solution, 1000 mL | 106.65 Online price 108.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|