eBioscience™ Essential Human T-Cell Phenotyping Kit
eBioscience™ Essential Human T-Cell Phenotyping Kit
Invitrogen™
eBioscience™ Essential Human T-Cell Phenotyping Kit
The Invitrogen eBioscience Essential Human T-Cell Phenotyping Kit is designed for flow cytometry-based identity testing of T-cells. It leverages high-qualityRead more
Have Questions?
Catalog NumberQuantity
A42923100 reactions
Catalog number A42923
Price (USD)
1,710.00
Each
In stock
Add to cart
Quantity:
100 reactions
Recurring order eligible. Learn more »
Price (USD)
1,710.00
Each
Add to cart
Ask our AI about this Product
The Invitrogen eBioscience Essential Human T-Cell Phenotyping Kit is designed for flow cytometry-based identity testing of T-cells. It leverages high-quality eBioscience antibodies and isotype controls and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.
Benefits include:
Proven performance―antibodies and supporting reagents designed for flow cytometry
Highly referenced antibodies―eBioscience portfolio contains trusted fluorochrome-conjugated antibodies and reagents for immunology and oncology research
Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
FormatKit
Quantity100 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Product LineeBioscience™
Product TypePhenotyping Kit
Unit SizeEach
Contents & Storage
  • CD3, CD4, CD8, CD62L, and CCR7 antibodies, store at 2–8°C
  • Isotype control, store at 2–8°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection