The Invitrogen eBioscience Essential Human Th1/Th17 Phenotyping Panel with Staining & Stimulation Reagents is designed for flow cytometry-based identity testing of Type 1 and Type 17 T-helper cells (Th1/Th17). It leverages high quality eBioscience antibodies, cell stimulation reagents (induces cytokine production), and fixation/permeabilization reagents (for intracellular markers IfN-g and IL-17) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.
Benefits include: • Proven performance―antibodies and supporting reagents designed for flow cytometry • Highly referenced antibodies―includes one of the most referenced clones for IL-17 • Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescent
Format
Kit
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line
eBioscience™
Product Type
Phenotyping Panel
Contents & Storage
CD4, Ifn-g, IL-17a antibodies, store at 2°C to 8°CC
Cell stimulating cocktail, store at -5 to -30°C
Protein transport cocktail, store at -5 to -30°C
Intracellular fixation & permealization reagents, store at 2°C to 8°CC