1x1 image pixel for data collection
Pierce™ β-Glucan Blocker
Pierce™ β-Glucan Blocker
Thermo Scientific™

Pierce™ β-Glucan Blocker

Pierce β-Glucan-Blocker is added to samples suspected of β-1,3-glucans contamination, preventing the occurrence of false positive results in endotoxin detectionRead more
Have Questions?
Catalog NumberQuantity
A438875 x 10 mL
Catalog number A43887
Price (USD)
54.75
Each
In stock
Add to cart
Quantity:
5 x 10 mL
Recurring order eligible. Learn more »
Price (USD)
54.75
Each
Add to cart
Ask our AI about this Product
Pierce β-Glucan-Blocker is added to samples suspected of β-1,3-glucans contamination, preventing the occurrence of false positive results in endotoxin detection (LAL) assays. β-1,3-glucans contamination may arise from the presence of yeast and cellulosic materials. β-1,3-glucans, in sufficient amounts, can trigger a LAL response not emanating from an endotoxin-mediated response. This blocker contains <0.005 EU/mL endotoxin.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Endotoxin Level<0.005 EU/mL
For Use With (Application)LAL Endotoxin Assay
FormatVial(s)
Quantity5 x 10 mL
Product TypeBeta-Glucans Blocker
SterilitySterile
Unit SizeEach
Contents & Storage
Store at 2 to 30°C.

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
0A13W00308Certificate of AnalysisJun 20, 2025A43887
0B33D00208Certificate of AnalysisMay 15, 2024A43887
0CA5V30808Certificate of AnalysisDec 07, 2023A43887
0D84T70608Certificate of AnalysisMar 09, 2023A43887
SPECSCertificate of AnalysisSep 20, 2022A43887
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection