Informational:Informational:Important: Please be informed that current sku number for CLA Identifiler Kit i.e. A44660 will be changing. Moving forward please use alternative sku – A65672. Note that no other changes (apart from the sku number) are applied to the product. Starting Jan 1st, 2025 – only A65672 will be available for sales.
Applied Biosystems™
CLA IdentiFiler™ Plus PCR Amplification Kit
Catalog number: A44660
Have Questions?
Applied Biosystems™
CLA IdentiFiler™ Plus PCR Amplification Kit
Catalog number: A44660
Have Questions?
Catalog Number
A44660
Unit Size
200 reactions
Price (USD)
7,070.00
Quantity
Catalog Number
Unit Size
Price (USD)
Quantity
A44660
200 reactions
7,070.00
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The CLA (Cell Line Authentication) IdentiFiler Plus PCR Amplification Kit simplifies the amplification step during authentication of cell lines. It includes the same primers and allelic ladder as the widely used AmpFLSTR Identifiler Plus kit and harnesses next-generation PCR amplification technology to help provide enhanced sensitivity, a cleaner baseline, improved performance with mixtures, and the ability to overcome high levels of PCR inhibition. The kit includes GeneScan 600 LIZ Size Standard v2.0, a five-dye-labeled, high-density size standard for the reproducible sizing of fragment analysis data.
Features of the CLA IdentiFiler Plus PCR Amplification Kit include: • 5-dye, 16-locus STR kit compatible with all cell line databasing loci standards • Improved version of legacy AmpFLSTR Identifiler Plus kit • Size standard included for ease of purchase • Compatible with SeqStudio, 3730/xL, and 3500/xL genetic analyzers
Cell line authentication using short tandem repeat (STR) analysis kits can be used both for human sample authentication (HSA) and mixed sample analysis (MSA). In HSA, there is a need to verify the DNA profile of the sample to check for sample cross-contamination or sample mix-up, or to perform a quality check. For MSA there is a need to deconvolute the presence of multiple contributors in a sample (chimerism) through identification of multiple DNA genotypes in the sample.
Specifications
Technique
SNP Genotyping (Fragment Analysis-Based), STR (Short Tandem Repeat) Analysis