GeneScan™ 600 LIZ™ dye Size Standard v2.0
GeneScan™ 600 LIZ™ dye Size Standard v2.0
Applied Biosystems™
GeneScan™ 600 LIZ™ dye Size Standard v2.0
The GeneScan™ 600 LIZ™ Size Standard v2.0 is a fifth dye-labeled high density size standard for the reproducible sizing ofRead more
Have Questions?
Catalog NumberQuantity
4408399800 reactions
Catalog number 4408399
Price (USD)
704.00
Each
In stock
Add to cart
Quantity:
800 reactions
Recurring order eligible. Learn more »
Price (USD)
704.00
Each
Add to cart
Ask our AI about this Product
The GeneScan™ 600 LIZ™ Size Standard v2.0 is a fifth dye-labeled high density size standard for the reproducible sizing of fragment analysis data. This updated version incorporates manufacturing process enhancements for improved lot to lot consistency and peak height balance compared to the existing GeneScan™ 600 LIZ™ Size Standard. This product is required for normalization on the 3500 Series Genetic Analyzers, and can also be used as a traditional size standard on all of the Applied Biosystems™ capillary electrophoresis systems. Use this size standard for fragments between 20 and 600 bp. The standard contains 36 LIZ™ dye-labeled, single-stranded DNA fragments. Since the standard is labeled with the fifth dye, users can genotype a greater number of markers in a given lane, compared to the four-dye system.
Size Fragments in the 20-600 Nucleotides Range
GeneScan™ 600 LIZ™ Size Standard v2.0 is designed for sizing DNA fragments in the 20-600 nucleotides range and provides 36 single-stranded labeled fragments of: 20, 40, 60, 80, 100, 114, 120, 140, 160, 180, 200, 214, 220, 240, 250, 260, 280, 300, 314, 320, 340, 360, 380, 400, 414, 420, 440, 460, 480, 500, 514, 520, 540, 560, 580 and 600. The sizing curve generated from fragments make the GeneScan™ 600 LIZ™ Size Standard v2.0 ideal for use for a variety of fragment analysis applications such as Microsatellites, Fragment Length Polymorphisms and Relative Fluorescent Quantitation. Each of the DNA fragments is labeled with the LIZ™ fluorophore which results in a single peak when run under denaturing conditions. With the 5th dye LIZ™ your marker fragments can be labeled with the dyes FAM™, VIC™ , NED™ or PET™.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer, 3500xL Dx Genetic Analyzer, SeqStudio Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Dx Genetic Analyzer
Label or DyeLIZ
Product TypeDye Size Standard V2.0
Quantity800 reactions
Shipping ConditionWet Ice
Product LineGeneScan™, LIZ™
Software TypeFragment Analysis
Starting MaterialDNA
Unit SizeEach
Contents & Storage
Each kit contains enough standard for 800 analyses.
Store in refrigerator (2–8°C).
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
What size standard do you recommend using with the TrueMark MSI Assay?
The GeneScan 600 LIZ dye Size standard v2.0 (Cat. No. 4408399) must be used with the TrueMark MSI Assay Analysis Software, and it has to be ordered separately.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
During fragment analysis, the larger peaks of my size standard are missing. Why?
If the larger peaks in the size standard are missing, it can also cause the sizing analysis to fail. This could be sample-related, where smaller ions and PCR products are preferentially injected into the capillaries and out-compete the injection of the larger products. To confirm, the size standard should be run without the sample.
If a size standard-only injection is performed, the larger size standard peaks may be missing due to slower migration of the sample. Slower migration can be due to some of the following:
-Degraded or expired polymer
- Old array or capillary
- Old or incorrect concentration of the buffer
- Old or expired HiDi Formamide
- Colder ambient temperature
- Oven temperature too low
If the issue persists, the instrument run time can be increased in order to allow for longer collection time of the largest size standard peaks. However, if the peak resolution is also broader than expected, this may suggest an instrument issue and a service call should be opened.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
During fragment analysis, I am missing the smaller fragments of my size standard. What happened?
If a size standard-only run is performed and is missing the smaller fragments, degradation of the size standard may have occurred. Please check the expiration date either on the box or the Certificate of Analysis (COA). The size standard should be stored at 4-8 degrees C. Freezing of the size standard will cause loss of the smaller products and may also result in dye breakdown. The size standard should be replaced if expired or stored improperly. If the smaller peaks are missing, they may also be masked by the primer peak. The easiest fix is to go to the GeneMapper Manager, Size Standard tab, select the size standard definition you are working with and click the “Save As” button at the bottom. Rename the new standard and edit it by deleting any size smaller than 50 bp. Click OK, then Done and change the Size Standard in your project and re-analyze the data.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
During fragment analysis, my sample peaks are really strong, but my size standard peaks are really low. Why is this?
If the sample has strong signal and the size standard is weak, the sample may be preferentially injecting into the capillaries. The sample may have a high salt concentration and/or robust amplification. To confirm, a size standard-only sample should be run to confirm the performance of the size standard alone. If the signal intensity improves, the sample may require desalting or less PCR product should be used. If the size standard-only sample still shows low signal, the issue may be with possible degradation of the size standard and it may need to be replaced.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
I am observing "failed sizing" in GeneMapper Software. Why?
In GeneMapper Software, “failed sizing” is a result of the software being unable to identify the size standard peaks as defined by the selected size Standard. Please confirm that the size standard used in the sample matches the size standard selected in GeneMapper Software. The Size Match Editor function in GeneMapper Software allows you to view the size standard peaks sizing assignments. In the Size Match Editor window, the size standard peak heights can be reviewed, the user can determine if all the peaks in the size standard are present, or if there are any extraneous peaks in the same color as the size standard that are present. The Size Match Editor can be accessed by highlighting the failed samples in the Samples tab, and then going to the Analysis pull down menu > Size Match Editor.
-Within the GeneMapper Software's Analysis Method, the Peak Detector tab contains the minimum peak height setting for each color (blue, green, yellow, red, purple, and orange). If the size standard peaks are below the minimum peak height setting on the Y-axis, typically 50, the software will ignore these peaks (this threshold is different for fragment analysis that occurs in the 3500 Data Collection software). Although this setting may be altered, it may be necessary to troubleshoot further before doing so. If the sample has strong signal and the size standard is weak, the sample may be preferentially injecting into the capillaries. The sample may have a high salt concentration and/or robust amplification. To confirm, a size standard-only sample should be run to confirm the performance of the size standard alone. If the signal intensity of the size standard improves, the sample may require desalting or less PCR product should be used. If the size standard-only sample still shows low signal, the issue may be possible degradation of the size standard or the Hi-Di Formamide and they may need to be replaced. It may also be a sign that the capillary array is old or clogged.
-If there are any extraneous peaks, this can be due to pull-up peaks from overloaded samples. Extraneous peaks in the same color as the size standard dye label may interfere with the sizing algorithm in the analysis software. The sample can be reduced to prevent the pull-up peak from overloaded samples. If the sample is not overloaded, please confirm that the fluorescent dye in use is part of the dye set selected. The spectral calibration may also need to be re-run.
- If the larger peaks in the size standard are missing, it can also cause the sizing analysis to fail. This could be sample-related where smaller ions and PCR products are preferentially injected into the capillaries and out-compete the injection of the larger products. To confirm, the size standard should be run without the sample.
- If a size standard-only injection is performed, the larger size standard peaks may be missing due to slower migration of the sample. Slower migration can be due to some of the following:
1.Degraded or expired polymer
2.Old array or capillary
3.Old or incorrect concentration of the buffer
4.Old or expired HiDi Formamide
5.Colder ambient temperature
6.Oven temperature too low
If the issue persists, the instrument run time can be increased in order to allow for longer collection time of the largest size standard peaks. However, if the peak resolution is also broader than expected, this may suggest an instrument issue and a service call should be opened.
- If the smaller peaks in the size standard are missing, they may be masked by the primer peak. The easiest fix is to go to the GeneMapper Manager, Size Standard tab, select the size standard definition you are working with and click the “Save As” button at the bottom. Rename the new standard and edit it by deleting any size smaller than 50 bp. Click OK, then Done and change the size standard in your project and re-analyze the data.
The Size Match Editor will allow overriding of the sample quality but it is important to confirm the size standard peaks are correctly identified. Otherwise, the peak sizes of the PCR products may be shifted. To override the sizing quality in the Size Match Editor, click on the “Override SQ” button on the top, middle of the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
3269036Certificate of AnalysisJun 27, 20254408399
3255540Certificate of AnalysisJun 06, 20254408399
3251873Certificate of AnalysisJun 02, 20254408399
3238831Certificate of AnalysisMay 14, 20254408399
3235916Certificate of AnalysisMay 09, 20254408399
5 results displayed, search above for a specific certificate
Safety Data Sheets
Limited Use Label Licenses (LULL)
'
Notice to Purchaser: This product is optimized for use in the DNA sequencing or fragment analysis methods covered by patents owned and/or controlled by Life Technologies Corporation (“LTC”). LTC does not convey any right or license under these patents, whether expressly, by implication, by estoppels, or otherwise, to the purchaser by the purchase of this product to use the DNA sequencing or fragment analysis methods. Notwithstanding the foregoing, a limited license to use the DNA sequencing or fragment analysis methods covered by such patents can be obtained for certain research and development activities (a) through the purchase of certain LTC reagents when such reagents are used in conjunction with an authorized LTC instrument, or (b) directly from LTC. For information on obtaining additional rights to practice the DNA sequencing or fragment analysis methods, please contact outlicensing@thermofisher.com, Licensing and Commercial Supply, Thermo Fisher Scientific, 5823 Newton Drive, Carlsbad, CA, 92008, United States.
'
Share catalog number, name or link