Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 5 x 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus DNA Ladder |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus DNA Ladder |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 2 x 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus DNA Ladder |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 5 x 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 2 x 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ Ultra Low Range DNA Ladder |
Gel Percentage | 4% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 10 to 500 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Compatible Ladders | E-Gel™ Ultra Low Range DNA Ladder |
Gel Percentage | 4% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 10 to 500 bp |
Stain Type | SYBR™ Safe |
Wells | 11-well |
Green Features | Less hazardous |
Quantity | 2 x 10 Gels |
Shipping Condition | Room Temperature |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Gel Percentage | Separation Range | Quantity | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A45205 | Each | 2% | 50 to 2000 bp | 5 x 10 Gels | 582.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A42100 | Each | 1% | 100 to 5000 bp | 10 Gels | 146.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A45202 | Each | 1% | 100 to 5000 bp | 2 x 10 Gels | 253.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A45203 | Each | 1% | 100 to 5000 bp | 5 x 10 Gels | 580.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A42135 | Each | 2% | 50 to 2000 bp | 10 Gels | 144.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A45204 | Each | 2% | 50 to 2000 bp | 2 x 10 Gels | 254.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A42136 | Each | 4% | 10 to 500 bp | 10 Gels | 183.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A45206 | Each | 4% | 10 to 500 bp | 2 x 10 Gels | 321.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features of E-Gel agarose gels include:
Safer option
Choose SYBR Safe DNA stained gels to minimize exposure to hazardous and mutagenic factors such as ethidium bromide and UV. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng of sample DNA per band and a separation time 2–3 fold faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a buffer-less ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.
Live monitoring of DNA migration
E-Gel agarose gels are pre-stained with SYBR Safe DNA gel stain which has an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, safe real-time monitoring of DNA migration is enabled without risk to the eyes or sample, as opposed to UV.
Easy access to the gel
The E-Gel cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for southern blot analysis.
Running and imaging devices
E-Gel agarose gels require the E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 11-well E-Gel agarose gels with SYBR Safe are available in 1%, 2%, and 4% gel percentages.