Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Cell and Tissue DNA Extraction Buffer

Catalog number: A45470
Have Questions?
Applied Biosystems™

MagMAX™ Cell and Tissue DNA Extraction Buffer

Catalog number: A45470
Have Questions?
Catalog Number
A45470
Unit Size
1000 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A454701000 preps
-
A45469100 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Cells, Tissue
For Use With (Application)
Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput Compatibility
Automated Protocols, High-throughput Compatible, Manual Protocols
No. of Reactions
1000 Preps
Product Line
MagMAX™
Product Type
Cell and Tissue DNA Extraction Buffer
Quantity
1000 Preps
Shipping Condition
Room Temperature
Target
Genomic DNA

Contents & Storage

600 mL Cell and Tissue DNA Extraction Buffer

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text