1x1 image pixel for data collection
AccuSEQ™ Real-Time PCR Detection Software, version 2.1.2
Product Image
Applied Biosystems™
AccuSEQ™ Real-Time PCR Detection Software, version 2.1.2
AccuSEQ Real-Time PCR Detection Software, version 2.1.2, is part of an integrated workflow for impurity and contaminant testing during biopharmaceuticalRead more
Have Questions?
Catalog NumberQuantity
A471301 each
Catalog number A47130
Price (USD)
21,230.00
Each
In stock
Add to cart
Quantity:
1 each
Price (USD)
21,230.00
Each
Add to cart
Ask our AI about this Product
AccuSEQ Real-Time PCR Detection Software, version 2.1.2, is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing using the SEQ family of real-time PCR assays, including MycoSEQ, resDNASEQ, and ProteinSEQ assays. The software is designed to work with the Applied Biosystems 7500 Fast Real-Time PCR Instrument with 0.1-mL, 96-well plates and Windows 10 operating system.
Features of AccuSEQ Real-Time PCR Detection Software include:
• Automated presence/absence calls for Mycoplasma detection
• Accurate quantitation of host cell line residual DNA
• Non-linear curve fitting of ProteinSEQ assay data for Protein A
• Single software platform for all current and future SEQ real-time PCR assays
• Security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance
Mycoplasma detection
For rapid and accurate Mycoplasma detection, AccuSEQ software offers a plug-in analysis module for automated presence/absence calls when using the MycoSEQ Mycoplasma Detection system. Automated analysis tools enable one-click processing of MycoSEQ assay data, helping deliver presence or absence calls within seconds of data collection being completed.
Residual DNA quantitation
For host cell residual DNA quantitation, a custom module with next-generation algorithms can be used with the resDNASEQ CHO Residual DNA Assay for accurate quantitation.
Protein quantitation
AccuSEQ software helps enable rapid and easy analysis of ProteinSEQ protein quantitation system data without the hassle of multiple exporting steps and reliance on third party software.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7500 Fast System
Product LineAccuSEQ™
Quantity1 each
Shipping ConditionRoom Temperature
Software TypeReal-Time PCR Software
Product TypeReal Time PCR Detection Software
Unit SizeEach
Contents & Storage
MycoSEQ, Probe, resDNASEQ
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection