Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit

Catalog number: A48310
Have Questions?
Applied Biosystems™

MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit

Catalog number: A48310
Have Questions?
Catalog Number
A48310
Unit Size
Up to 2,000 preps
Price (USD)
Quantity
Catalog Number
Unit Size
Up to 200 preps
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A48310Up to 2,000 preps
A42352Up to 200 preps
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM). The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing.

Features of the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit include:
• Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
• Fast procedure allows for 96 samples to be processed in under 30 minutes
• Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of biofluids or transfer media
• No need for carrier RNA
• Elution volumes ranging from 50 to 100 μL

Maximum sample input in a 96-deep well or 24-deep well plate format
The optimized reagents included in the MagMAX Viral/Pathogen kit allow you to maximize the amount of sample input, thereby increasing the amount of RNA and/or DNA recovered. Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate. For larger sample volumes, protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate.

Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.

The Dynabeads MyOne Silane contained in the MagMAX Viral/Pathogen Total Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with a highly specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery viral RNA and DNA present at low concentrations in body fluids. Once captured, these nucleic acids can then be eluted in 50 μL of elution buffer for use in downstream applications.

Specifications

Format
96 Deep-well Plate, 24 Deep-well Plate
Purification Time
30 min.
Product Line
MagMAX™
Quantity
Up to 2000 Preps
Shipping Condition
Room Temperature

Contents & Storage

• 550 mL Binding Solution, store at room temperature
• 1000 mL Wash Solution, store at room temperature
• 100 mL Elution Solution, store at room temperature
• 10 mL Proteinase K, store at room temperature
• 20 mL DNA/RNA Binding Beads, store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text