Human Plasma-Like Medium (HPLM)
Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
Human Plasma-Like Medium (HPLM)
Gibco™

Human Plasma-Like Medium (HPLM)

Gibco Human Plasma-like Medium (HPLM) is designed to resemble the natural cellular environment found in the body, mimicking the metabolic profile of human plasma.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A4899101500 mL
A489910210 x 500 mL
Catalog number A4899101
Price (EUR)
70,25
Each
-
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Price (EUR)
70,25
Each
Add to cart
Ask our AI about this Product

Gibco Human Plasma-like Medium (HPLM) is designed to resemble the natural cellular environment found in the body, mimicking the metabolic profile of human plasma.

  • Physiologically relevant—designed to resemble the natural cellular environment found in the body; formulated with over than 60 polar metabolites and plasma-like salt concentrations
  • Peer reviewed—extensive publication of research that used HPLM
  • Easy to use—direct replacement for conventional media when supplemented with FBS

The widely used classic synthetic cell culture media, including MEM, DMEM, RPMI 1640, and DMEM/F-12 contain glucose, amino acids, vitamins, and salts at concentrations that often do not reflect those found in human plasma. These media also lack additional plasma components needed to mimic the metabolic profile of human plasma. When studying cancer and other diseases, results with more physiological relevance will enable researchers to improve their understanding of human function and illness.

The HPLM solution

HPLM contains more than 60 polar metabolites such as amino acids, nucleic acids, sugars, and small organic acids at concentrations found in human plasma. The salt concentrations in HPLM mimic the salt concentrations found in human plasma. In resembling the natural cellular environment found in the body, HPLM provides researchers with the ability to study the impact of physiologically relevant cell media on their specific applications.

When supplemented with fetal bovine serum (FBS), HPLM is capable of supporting cell growth and viability comparable to those of conventional basal media supplemented with FBS. For most cell lines, adaptation is not required to transition from conventional medium to HPLM.

The complete HPLM formulation is available (hyperlink to technical-resource page with complete HPLM formulation).

cGMP manufacturing and quality system

HPLM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications
Cell TypeMammalian Cells
For Use With (Application)Mammalian Cell Culture
Product TypeHuman Plasma-Like Medium
Quantity500 mL
Shelf Life12 Months
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo HEPES
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3185068Certificate of AnalysisMay 29, 2025A4899101
3185061Certificate of AnalysisMay 29, 2025A4899101
3185059Certificate of AnalysisMay 15, 2025A4899101
2996700Certificate of AnalysisMar 13, 2025A4899101
2899744Certificate of AnalysisMar 13, 2025A4899101
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Yes, Gibco HPLM is animal origin-free (AOF).

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Yes, Gibco HPLM is manufactured in a cGMP facility.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

There are several publications using HPLM. A list of select publications using HPLM can be found on the HPLM web page: https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/classical-media/human-plasma-like-medium.html

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

The complete formulation of Gibco Human Plasma-Like Medium (HPLM) can be found here .

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Gibco HPLM is not a BenchStable media and should not be stored at room temperature. Recommended storage for Gibco HPLM is at 2-8 degrees C, protected from light.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Share catalog number, name or link

1x1 image pixel for data collection