Gibco™

Human Plasma-Like Medium (HPLM)

Catalog number: A4899102
Have Questions?
Gibco™

Human Plasma-Like Medium (HPLM)

Catalog number: A4899102
Have Questions?
Catalog Number
A4899102
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A489910210 x 500 mL
-
A4899101500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Human Plasma-like Medium (HPLM) is designed to resemble the natural cellular environment found in the body, mimicking the metabolic profile of human plasma.

Physiologically relevant—designed to resemble the natural cellular environment found in the body; formulated with over than 60 polar metabolites and plasma-like salt concentrations
Peer reviewed—extensive publication of research that used HPLM
Easy to use—direct replacement for conventional media when supplemented with FBS

The widely used classic synthetic cell culture media, including MEM, DMEM, RPMI 1640, and DMEM/F-12 contain glucose, amino acids, vitamins, and salts at concentrations that often do not reflect those found in human plasma. These media also lack additional plasma components needed to mimic the metabolic profile of human plasma. When studying cancer and other diseases, results with more physiological relevance will enable researchers to improve their understanding of human function and illness.

The HPLM solution
HPLM contains more than 60 polar metabolites such as amino acids, nucleic acids, sugars, and small organic acids at concentrations found in human plasma. The salt concentrations in HPLM mimic the salt concentrations found in human plasma. In resembling the natural cellular environment found in the body, HPLM provides researchers with the ability to study the impact of physiologically relevant cell media on their specific applications.

When supplemented with fetal bovine serum (FBS), HPLM is capable of supporting cell growth and viability comparable to those of conventional basal media supplemented with FBS. For most cell lines, adaptation is not required to transition from conventional medium to HPLM.

The complete HPLM formulation is available (hyperlink to technical-resource page with complete HPLM formulation).

cGMP manufacturing and quality system
HPLM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Cell Type
Mammalian Cells
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Human Plasma-Like Medium
Serum Level
Standard Serum Supplementation
Shelf Life
12 Months
Sterility
Sterile
With Additives
Glutamine, Phenol Red
Without Additives
No HEPES
For Use With (Application)
Mammalian Cell Culture
Quantity
10 x 500 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text