AAV-MAX Transfection Kit
AAV-MAX Transfection Kit
Gibco™

AAV-MAX Transfection Kit

The Gibco AAV-MAX Transfection Kit is an essential component of the AAV-MAX Helper-Free AAV Production System. It is formulated forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A505161 kit for 10 L of production
A505151 kit for 1 L of production
Catalog number A50516
Price (USD)
8,680.65
Online Exclusive
9,470.00
Save 789.35 (8%)
Each
-
Add to cart
Quantity:
1 kit for 10 L of production
Price (USD)
8,680.65
Online Exclusive
9,470.00
Save 789.35 (8%)
Each
Add to cart
Ask our AI about this Product
The Gibco AAV-MAX Transfection Kit is an essential component of the AAV-MAX Helper-Free AAV Production System. It is formulated for the transient transfection of Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from Viral Production Cells 2.0 when cultured in Viral Production Medium.

Features of the AAV-MAX Transfection Kit include:
• Components work synergistically within the AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
• AAV-MAX transfection reagent—cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
• AAV-MAX booster—chemically defined component that works with transfection reagent to achieve high transfection efficiency
• AAV-MAX enhancer—chemically defined component that enhances the production of AAV particles
• Robust transfection protocols are adaptable to large scale transfection
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineHEK293
ClassificationAnimal Origin-free
Culture TypeSuspension Cell Culture
DescriptionSerum-free, animal origin-free, chemically defined
Expression SystemMammalian
For Use With (Application)AAV Production
For Use With (Equipment)AAV-MAX System
Includes100 mL Gibco AAV-MAX Enhancer, 60 mL Gibco AAV-MAX Transfection Reagent, 30 mL Gibco AAV-MAX Supplement
Manufacturing QualityISO 13485, ISO 9001 certified
Packaging TypeBottle
Product LineAAV-MAX
Product TypeAAV production transfection Kit
Quantity1 kit for 10 L of production
Reagent TypeTransfection kit
Shelf Life12 months from date of manufacture
Shipping ConditionWet Ice
SterilitySterile-filtered
Cell TypeEstablished Cell Lines
FormatKit
Sample TypePlasmid DNA
Transfection TechniqueLipid-based Transfection, Transient Transfection
Unit SizeEach
Contents & Storage
• 60 mL AAV-MAX Trasfection Reagent
• 30 mL AAV-MAX Transfection Booster
• 100 mL AAV-MAX Enhancer

Reagents should be stored at 2–8°C. Do not freeze.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3160255Certificate of AnalysisJun 24, 2025A50515
3131719Certificate of AnalysisMay 06, 2025A50516
3124763Certificate of AnalysisApr 09, 2025A50515
3047160Certificate of AnalysisFeb 07, 2025A50515
3047159Certificate of AnalysisJan 31, 2025A50516
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The AAV-MAX Enhancer increases AAV vector production in Viral Production Cells (VPCs) 2.0. The identity and mechanism of action of the AAV-MAX Enhancer are proprietary.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

The AAV-MAX Transfection Booster enhances the efficiency of co-transfection of multiple plasmids into high-density Viral Production Cells (VPCs) 2.0. The identity and mechanism of action of the transfection booster are proprietary.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

The AAV-MAX Transfection Reagent, AAV-MAX Tranfection Booster, and AAV-MAX Enhancer are specifically designed to work synergistically with the Viral Production Medium and Viral Production Cells 2.0 to provide maximum titers. Mixing and matching of reagents is not recommended, as this will decrease the overall performance.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

The AAV-MAX transfection protocol is scalable from 24 deep-well plates to shake flasks to bioreactors. The user guide provides guidance on this.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Yes, these are different, and each is optimized for its intended use.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

Share catalog number, name or link

1x1 image pixel for data collection