Gibco™

Viral Production Cells 2.0

Catalog number: A51218
Have Questions?
Gibco™

Viral Production Cells 2.0

Catalog number: A51218
Have Questions?
Catalog Number
A51218
Unit Size
6 vials
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 vial
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A512186 vials
-
A497841 vial
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F parental cell line and a core component of the AAV-MAX Helper-Free AAV Production System. The cells have been adapted to the chemically defined Viral Production Medium and are maintained in suspension culture in that medium. Viral Production Cells 2.0 are highly transfectable using the AAV-MAX Transfection Kit for adeno-associated virus (AAV) production.

Features of the Viral Production Cells 2.0 include:
• Clonal, 293F-derived, high-producer cell line
• Absence of SV40 large T antigen
• Optimized for high-density, suspension culture (>12 million cells/mL) in chemically defined medium
• Enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
• Robust scalability and passage stability

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Viral Production Medium and 10% DMSO. Cells should be thawed directly into Viral Production Medium supplemented with 4 mM GlutaMAX Supplement (Cat. No. 35050038) for optimal performance.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell State
Frozen
Concentration
1e7 cells/mL
Cell Line
Viral Production Cells 2.0
Cell Type
Kidney (Embryonic)
Culture Type
Suspension Cell Culture
Form
Frozen
Species
Human
Format
6 vials
For Use With (Application)
AAV Production
Media Recommendation
Gibco Viral Production Medium
No. of Cells
1 x 107 cells
Product Line
AAV-MAX
Product Type
Viral Production Cells
Quantity
1 mL
Shipping Condition
dry ice for domestic shipment; liquid nitrogen for international shipment
Source
Human

Contents & Storage

6 cryovials, store in vapor phase liquid nitrogen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text