Absolute Q™ DNA Digital PCR Master Mix (5X)
Absolute Q™ DNA Digital PCR Master Mix (5X)
Applied Biosystems™

Absolute Q™ DNA Digital PCR Master Mix (5X)

Applied Biosystems Absolute Q DNA Digital PCR Master Mix (5X) is optimized for use with the QuantStudio Absolute Q DigitalRead more
Have Questions?
Catalog NumberQuantity
A52490200 reactions
Catalog number A52490
Price (USD)
363.00
Each
-
Add to cart
Quantity:
200 reactions
Recurring order eligible. Learn more »
Price (USD)
363.00
Each
Add to cart
Ask our AI about this Product
Applied Biosystems Absolute Q DNA Digital PCR Master Mix (5X) is optimized for use with the QuantStudio Absolute Q Digital PCR System and Applied Biosystems digital PCR (dPCR) assays in a simple workflow with minimal processing steps. The 5X formulation enables analysis of higher sample volumes and delivers accurate quantification of DNA targets without using a standard curve.

Features of Absolute Q DNA Digital PCR Master Mix include:
Fast time-to-results—5 minutes of hands-on time and results in ∼90 minutes
Simple workflow—set up the reaction like qPCR, then load and run the plate in a single instrument
High sensitivity—ability to detect down to 0.1% mutant allele frequency
Optimized for multiplexing—detect up to four targets in a single reaction

Simple and efficient workflow
The Absolute Q system simplifies the entire dPCR workflow. Prepare the dPCR reaction mix as you would for qPCR, pipette the reaction mix and isolation buffer into the microfluidic array plate (MAP), and place the plate in the QuantStudio Absolute Q Digital PCR System. Compartmentalization, thermal cycling, and data acquisition of up to four targets occur in a single instrument.

High sensitivity
Absolute Q DNA Digital PCR Master Mix (5X) enables analysis of >95% of the loaded reaction and consistently generates >20,000 microreactions, which maximizes sensitivity. Absolute Q DNA Digital PCR Master Mix (5X) can detect down to 0.1% mutant allele frequency when paired with Absolute Q liquid biopsy dPCR assays.

Maximum specificity and accuracy
Absolute Q DNA Digital PCR Master Mix (5X) is formulated with ROX dye for accurate micro-chamber analysis and with hot-start Taq DNA polymerase and dUTP/UDG to deliver high specificity and carryover contamination control.

DNA methylation analysis
For DNA methylation analysis, use Absolute Q 1-step RT-dPCR Master Mix (4X), which does not contain UNG.

Related products
Absolute Q Liquid Biopsy dPCR assays
Absolute Q Viral Titer dPCR assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5X
For Use With (Equipment)QuantStudio Absolute Q Digital PCR System
No. of Reactions200 reactions
PolymeraseTaq DNA Polymerase
Product LineAbsolute Q
Product TypeDigital PCR Master Mix
Quantity200 reactions
Sample TypeDNA, cDNA
Shipping ConditionDry Ice
Sufficient For200 Reactions
Volume360 μL
Detection MethodPrimer-probe
For Use With (Application)Digital PCR (dPCR)
FormLiquid
PCR MethodDigital PCR
Reaction SpeedFast
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3219747Certificate of AnalysisMay 30, 2025A52490
3219743Certificate of AnalysisApr 18, 2025A52490
3193162Certificate of AnalysisApr 07, 2025A52490
3174748Certificate of AnalysisMar 20, 2025A52490
3159431Certificate of AnalysisFeb 11, 2025A52490
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Yes, ROX is the passive reference dye for QC (Quality Control).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Share catalog number, name or link

1x1 image pixel for data collection