Invitrogen™

Colorimetric ReadiLAMP™ Kit, SARS-CoV-2

Catalog number: A52544
Have Questions?
Invitrogen™

Colorimetric ReadiLAMP™ Kit, SARS-CoV-2

Catalog number: A52544
Have Questions?
Catalog Number
A52544
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A525441000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The Invitrogen Colorimetric ReadiLAMP Kit for SARS-CoV-2 is a loop-mediated isothermal amplification (LAMP)-based solution for rapid SARS-CoV-2 detection in saliva and nasopharyngeal/nasal swab samples. This kit offers two protocol options. The first provides a workflow for RNA purification followed by a colorimetric LAMP assay. (For robust SARS-CoV-2 RNA isolation, the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) or PureLink Viral RNA/DNA Mini Kit (Cat. No. 12280050) are recommended.) The second protocol consists of a simple SARS-CoV-2 viral lysis method for compatible sample types followed by a colorimetric LAMP assay. This second workflow does not require nucleic acid isolation or complex sample preparation; instead it directly incorporates the saliva or nasopharyngeal/nasal swab samples as LAMP assay input.

Features of the Colorimetric ReadiLAMP Kit for SARS-CoV-2 include:
Simplicity—detection is fast, simple, and intuitive; LAMP reaction changes color from purple to blue if SARS target is present; no instrumentation required, except for a 65°C heating block
Sensitivity—input of up to 50% volume of purified RNA per LAMP reaction, enabling maximal sensitivity in SARS-CoV-2 detection
    --Direct LAMP: 100–500 copies per reaction (25 μL)
    --Purified RNA-based LAMP: 20–100 copies per reaction (25 μL)
Speed
    --Direct LAMP: ∼35 minutes using crude saliva or swabs in non-serum-containing transport medium
    --Purified RNA-based LAMP: ∼ 60 minutes (∼30 minutes RNA purification, 30 minutes LAMP assay)
Throughput—process 1–96 samples at a time, enabling processing of more samples with same personnel headcount, using individual tubes, tube strips, or 96-well/384-well plates

Specifications

Bead Type
Kit does not contain beads. Recommending the use of RNA extraction MVP II kit which contains magnetic beads
Detection Limit
<250 copies/25 μL (Direct LAMP)
<100 copies/25 μL (Purified RNA-based LAMP)
Color
During reaction color changes from purple to blue
Detection Method
Colorimetric readout
Sample Type
Saliva or Nasopharyngeal/Nasal Swab
Enzyme
SuperScript IV & proprietary Bst-derived DNA polymerase
For Use With (Application)
SARS-CoV-2 surveilance
Formulation
Liquid. 5 components included in the kit.
For Use With (Equipment)
Heating block
Includes
LAMP Buffer (5x), Enzyme Mix (10x), SARS-CoV-2 primer mix (5x), Human Control primer mix (5x), SARS-CoV-2 control RNA
Key Functions
Detects SARS-CoV-2 presence
Sample Volume
Range of sample inputs
Tested For
SARS-CoV-2 presence
No. of Reactions
1000
Unit Size
1000 reactions

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text