Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | TMTpro™ 18plex Label Reagents in stabilized solution, 96-well plate, 4 x 50 μg |
Formulation | Liquid in stabilized DMSO solution |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Isobaric Label Reagent Set |
Product Line | TMT™ |
Label or Dye | Tandem Mass Tag™ (TMT) Labels |
Starting Material | Cell Lysate |
Final Product Type | Protein PEPTIDE |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Labeling |
Quantity | 72 Reactions |
Form | Liquid |
Unit Size | 4 x 50 μg (per tag) |
Description | TMTpro™ 18-plex Label Reagent Set, 5 mg |
Product Type | Isobaric Label Reagent Set |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide labeling and quantitation |
Quantity | 5 mg |
Unit Size | 1 x 5 mg |
Description | TMTpro-134C and TMTpro-135N Label Reagents, 5 mg |
Product Type | Isobaric Label Reagent Set |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide labeling and quantitation |
Quantity | 5 mg |
Unit Size | 1 x 5 mg |
Description | TMTpro™ 18-plex Label Reagent Set, 6 x 0.5 mg |
Product Type | Isobaric Label Reagent Set |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide labeling and quantitation |
Quantity | 6 x 0.5 mg |
Unit Size | 6 x 0.5 mg |
Description | TMTpro-134C and TMTpro-135N Label Reagents, 6 x 0.5 mg |
Product Type | Isobaric Label Reagent Set |
Starting Material | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Peptide labeling and quantitation |
Quantity | 6 x 0.5 mg |
Unit Size | 6 x 0.5 mg |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A58335 | 4 x 50 μg (per tag) | TMTpro™ 18plex Label Reagents in stabilized solution, 96-well plate, 4 x 50 μg | 515.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A52045 | 1 x 5 mg | TMTpro™ 18-plex Label Reagent Set, 5 mg | 9,350.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A52046 | 1 x 5 mg | TMTpro-134C and TMTpro-135N Label Reagents, 5 mg | 1,058.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A52047 | 6 x 0.5 mg | TMTpro™ 18-plex Label Reagent Set, 6 x 0.5 mg | 8,470.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A52048 | 6 x 0.5 mg | TMTpro-134C and TMTpro-135N Label Reagents, 6 x 0.5 mg | 942.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The structures of the TMTpro and TMT tags are similar in being isobaric and amine reactive, but the TMTpro tag has a longer spacer region and isobutyl proline mass reporter. After MS/MS fragmentation, each TMTpro 18plex tag generates a unique reporter mass (e.g., TMTpro 126-135 Da) in the low-mass region of the high-resolution MS/MS spectrum that is used for relative quantitation of protein expression levels.
The TMT pro134C and TMTpro135N Label Reagents are identical in composition and structure to the other TMTpro 16plex tags, but are not isobaric. Addition of the TMTpro 134C and 135N tags create a 17th and 18th channel for relative quantitation using high resolution Orbitrap MS instruments. These tags are available separately (Cat. No. A52046, A52048), as part of the TMTpro 18plex Label Reagent Set, or as part of the complete 18plex set of reagents in a 96-well plate format.
Features of the TMTpro 18plex Label Reagent Set include:
• Multiplex—concurrent MS analysis of up to 18 samples derived from cells, tissues, or biological fluids
• Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
• Efficient—amine-reactive, NHS ester-activated reagents ensure greater than 95% labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
• Optimized for use—with high resolution Thermo Scientific MS/MS Orbitrap platforms, analysis fully supported by Proteome Discoverer 2.3
• Convenient size—provided in a ready-to-use, single-use format to ensure optimal stability and performance
For high resolution analysis of TMTpro-labeled peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES902 or ES903). When combined with the industry-leading, high resolution Orbitrap instruments and software, TMTpro reagents provide an integrated total solution for quantitative protein expression analysis.
Sufficient for: Four 18-plex (4 x 18-way) experiments
• TMTpro-126 Label Reagent, 4 x 50 μg
• TMTpro-127 N Label Reagent, 4 x 50 μg
• TMTpro-127 C Label Reagent, 4 x 50 μg
• TMTpro-128 N Label Reagent, 4 x 50 μg
• TMTpro-128 C Label Reagent, 4 x 50 μg
• TMTpro-129 N Label Reagent, 4 x 50 μg
• TMTpro-129 C Label Reagent, 4 x 50 μg
• TMTpro-130 N Label Reagent, 4 x 50 μg
• TMTpro-130 C Label Reagent, 4 x 50 μg
• TMTpro-131 N Label Reagent, 4 x 50 μg
• TMTpro-131 C Label Reagent, 4 x 50 μg
• TMTpro-132 N Label Reagent, 4 x 50 μg
• TMTpro-132 C Label Reagent, 4 x 50 μg
• TMTpro-133 N Label Reagent, 4 x 50 μg
• TMTpro-133 C Label Reagent, 4 x 50 μg
• TMTpro-134 N Label Reagent, 4 x 50 μg
• TMTpro-134 C Label Reagent, 4 x 50 μg
• TMTpro-135 N Label Reagent, 4 x 50 μg
Store at -20°C.