Gibco™

Residual Protein TrypLE™ ELISA Kit

Catalog number: A6533103
Have Questions?
Gibco™

Residual Protein TrypLE™ ELISA Kit

Catalog number: A6533103
Have Questions?
Catalog Number
A6533103
Unit Size
10 kits
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 kits
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A653310310 kits
-
A65331011 kit
-
A65331025 kits
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The Residual Protein TrypLE ELISA Kit is a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) for the quantitative detection of TrypLE enzyme. The kit is compatible with all forms of TrypLE enzyme, including TrypLE Select, TrypLE Express, and CTS TrypLE Select enzymes.

TrypLE enzyme is an animal-origin-free recombinant alternative to porcine trypsin. It is used for the dissociation of attachment-dependent cell lines. TrypLE enzyme dissociates cells cultured both in serum-free and serum-supplemented systems. Due to the unique biological sequence of TrypLE enzyme, traditional trypsin ELISA kits are ineffective at binding and detecting residual levels of TrypLE enzyme.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Assay Range
0.78 to 50 ng/mL
Assay Sensitivity
0.032 ng/mL
Conjugate
Horseradish Peroxidase (HRP)
Immunoassay Kit Format
Sandwich ELISA Kit
For Use With (Equipment)
Colorimetric Microplate Reader
Interassay CV
6.20%
Intraassay CV
7.20%
Label or Dye
Horseradish Peroxidase (HRP)
Protein Aliases
TrypLE, R-Protease
Quantity
960 Tests
Shipping Condition
Wet Ice
Target
TrypLE (R-Protease) concentration
Total Assay Time
1.5 hr.
Format
Wash steps required

Contents & Storage

• Pre-coated 96-well plate
• Standard assay buffer concentrate
• HRP-conjugated detection antibody
• Wash buffer concentrate
• Chromogen stop solution
• Adhesive plate covers

Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text