Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Select the Thermo Scientific™ Orion™ AQUAfast AQ3700 Colorimeter to manage your water and wastewater analysis needs. This instrument has over 90 pre-programmed methods for common test parameters with automatic selection of 6 LED wavelengths and countdown timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank. This IP67 waterproof meter records 1,000 data points, offers a long battery life of about 26 hours continuous use or 3,500 tests, and multi-language user interface in English, French, German, Italian, Polish, Portuguese and Spanish.
Certifications/Compliance | FCC Part 15; CE |
Type | Meter Verification Check Standards Kit |
For Use With (Equipment) | AQ3700 Colorimeter |
Storage Requirements | 5°C to 40°C |
Unit Size | Each |
Certifications/Compliance | FCC Part 15; CE |
Light Source | 6 x LED |
Type | Replacement Adapter, 16 mm |
For Use With (Equipment) | AQ3700 Colorimeter |
Height (Metric) | 13 mm |
Length (English) | 0.5 in. |
Storage Requirements | 5°C to 40°C |
Unit Size | Each |
Type | Sealing Ring for 24 mm Vial |
For Use With (Equipment) | AQ3700 Colorimeter |
Storage Requirements | 5°C to 40°C |
Unit Size | Case of 12 |
Accuracy | 2% Full scale |
Backlight Option | Yes |
Certifications/Compliance | FCC Part 15; CE |
Cuvette Details | Round 13 mm, 16 mm, 24 mm & 16 mm COD Digestion Tubes |
Battery Type | AA |
Display | Graphic |
Languages | English, French, German, Spanish, Italian |
Light Source | 6 x LED |
Product Line | AQUAfast |
Type | Colorimeter |
Wavelength Range | 430, 530, 560, 580, 610, 660 nm |
Interface | Internal RJ45 port |
Keypad | Numeric keys, ON/OFF, Up, Down, Enter, Timer, Zero, Test, Cal, Mode, Store, Shift, plus 3 Function keys |
Outputs | IRDA |
Reagent Form | Powder, tablet, liquid, reagent tubes, digestion tubes. Color, Turbidity and Suspended Solids tests do not require reagents |
Resolution | ±0.005 A |
Sample Size | 10 mL |
Storage Requirements | 5°C to 40°C |
Test Method | Up to 90 and 10 user defined programmed methods |
Unit Size | Each |
Certifications/Compliance | FCC Part 15; CE |
Type | Replacement Adapter, 13 mm |
For Use With (Equipment) | AQ3700 Colorimeter |
Height (English) | 0.6 in. |
Height (Metric) | 16 mm |
Storage Requirements | 5°C to 40°C |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AC3VSK | Each | Meter Verification Check Standards Kit | 322.50 Online price 375.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC37A13 | Each | Replacement Adapter, 16 mm | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC3SR24 | Case of 12 | Sealing Ring for 24 mm Vial | 36.54 Online price 40.60 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AQ3700 | Each | Colorimeter | 1,395.00 Online price 1,550.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC37A16 | Each | Replacement Adapter, 13 mm | 108.90 Online price 121.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Requires:
Recommended for: