Orion™ AQ3700 AQUAfast™ Colorimeter
Orion™ AQ3700 AQUAfast™ Colorimeter
Thermo Scientific™

Orion™ AQ3700 AQUAfast™ Colorimeter

Select this multiparameter colorimeter to manage your water and wastewater analysis using over 90 preprogrammed methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
AQ3700Colorimeter
AC37A13Replacement Adapter, 16 mm
AC3VSKMeter Verific Check Stds Kit
AC37A16Replacement Adapter, 13 mm
Catalog number AQ3700
Price (USD)
1,539.00
Online Exclusive
1,710.00
Save 171.00 (10%)
Each
In stock
Add to cart
Type:
Colorimeter
Recurring order eligible. Learn more »
Price (USD)
1,539.00
Online Exclusive
1,710.00
Save 171.00 (10%)
Each
Add to cart
Ask our AI about this Product

Select the Thermo Scientific™ Orion™ AQUAfast AQ3700 Colorimeter to manage your water and wastewater analysis needs. This instrument has over 90 pre-programmed methods for common test parameters with automatic selection of 6 LED wavelengths and countdown timer. Methods include total nitrogen and total phosphate nutrient testing or create up to 10 custom methods using 2-14 standards and blank. This IP67 waterproof meter records 1,000 data points, offers a long battery life of about 26 hours continuous use or 3,500 tests, and multi-language user interface in English, French, German, Italian, Polish, Portuguese and Spanish.

Features:

  • Wide range of disinfectants in various formats to broaden our offering to a variety of industries
  • Large graphic display with step-by-step user instructions
  • Automatic countdown to measurement
  • Consistent results from shift to shift
  • Selectable languages of English, French, German, Italian, Spanish, Portuguese (Brazilian), and Polish with other languages uploadable via RJ 45 connection

Requires:

  • 4 AA batteries

Recommended for:

  • Wastewater
  • Drinking water
  • Nutrient monitoring
  • Environmental testing
  • Food and beverage
  • Industrial applications
Note: For the most accurate results, use only Thermo Scientific Orion reagents.

 

 

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy2% Full scale
Backlight OptionYes
Certifications/ComplianceFCC Part 15; CE
Cuvette DetailsRound 13 mm, 16 mm, 24 mm & 16 mm COD Digestion Tubes
IncludesAQ3700 colorimeter, 4 batteries (Type AA/LR 6); Round 24 mm vials with caps; Round 16 mm vials with caps; 16 mm vial adapter; 13 mm vial adapter; Vial cleaning brush; Stirring rod
InterfaceInternal RJ45 port
KeypadNumeric keys, ON/OFF, Up, Down, Enter, Timer, Zero, Test, Cal, Mode, Store, Shift, plus 3 Function keys
OutputsIRDA
Reagent FormPowder, tablet, liquid, reagent tubes, digestion tubes. Color, Turbidity and Suspended Solids tests do not require reagents
Resolution±0.005 A
Sample Size10 mL
Storage Requirements5°C to 40°C
Test MethodUp to 90 and 10 user defined programmed methods
Battery TypeAA
DisplayGraphic
LanguagesEnglish, French, German, Spanish, Italian
Light Source6 x LED
Product LineAQUAfast
TypeColorimeter
Wavelength Range430, 530, 560, 580, 610, 660 nm
Unit SizeEach

Additional resources

We are continuously improving our website. While we work to include all support materials in one location, you’ll find the following information here:

Certificates of Analysis
SDS

Preprogrammed Methods Software Download

Warranty registration form

Register your Orion, ROSS, AQUAfast and AQUASensors products

Warranty registration

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Share catalog number, name or link

1x1 image pixel for data collection