Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Buffer Kit, RNase-free

Catalog number: AM9010
Have Questions?
Invitrogen™

Buffer Kit, RNase-free

Catalog number: AM9010
Have Questions?
Catalog Number
AM9010
Unit Size
1 set
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM90101 set
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ambion™ Buffer Kit provides high quality, RNase free buffers for critical RNA analysis procedures. The kit consists of one 100 mL bottle of each of the following solutions: 1 M Tris pH 7.0, 1 M Tris pH 8.0, 5 M NaCl, 1 M MgCl2, 0.5 M EDTA pH 8.0, 2 M KCl, 5 M Ammonium Acetate, and DEPC-treated H2O. Ammonium Acetate is sterile filtered. All other solutions are autoclaved post-packaging to ensure sterility. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Format
Kit
Kit Type
Buffer Kit
Product Line
Ambion™
Purity or Quality Grade
DNase-Free, RNase-Free
Shipping Condition
Room Temperature
For Use With (Application)
miRNA analysis
Volume
8 x 100 mL

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text