Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

EDTA (0.5 M), pH 8.0, RNase-free

Catalog number: AM9262
Have Questions?
Invitrogen™

EDTA (0.5 M), pH 8.0, RNase-free

Catalog number: AM9262
Have Questions?
Catalog Number
AM9262
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM92621 L
-
AM9260G100 mL
-
AM9261500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Ambion Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name or Material
EDTA
Concentration
0.5 M
Packaging Type
Bottle
pH
8
Product Line
Ambion™
Purity
Molecular Biology Grade
Quantity
1 L
Shipping Condition
Room Temperature
Form
Liquid

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text