EVOS™ Power Cord, Type D (India/South Africa)
EVOS™ Power Cord, Type D (India/South Africa)
Thermo Scientific™
EVOS™ Power Cord, Type D (India/South Africa)
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries withRead more
Have Questions?
Catalog NumberQuantity
AMEP47081 Ea.
Catalog number AMEP4708
Price (MXN)
-
Quantity:
1 Ea.
Ask our AI about this Product
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type D power plugs. The Type D electrical plug is used in parts of Africa, Asia, and South America.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Connector TypesType D (Electrical Plug), Type D (Power Plug)
For Use With (Equipment)EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Quantity1 Ea.
Product LineEVOS
TypePower Cord
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Other products to consider
Share catalog number, name or link