Invitrogen™

20X Bolt™ MOPS SDS Running Buffer

Catalog number: B000102
Have Questions?
Invitrogen™

20X Bolt™ MOPS SDS Running Buffer

Catalog number: B000102
Have Questions?
Catalog Number
B000102
Unit Size
5 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B0001025 L
-
B0001500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Bolt MOPS SDS Running Buffer (20X) is formulated for running proteins on Bolt Bis-Tris Plus gels. It is recommended for separating medium- to large-sized proteins.

Use the right buffer to optimize protein separations
Bolt MES SDS Running Buffer and Bolt MOPS SDS Running Buffer can both be used with Bolt Bis-Tris Plus gels. MES has a lower pKa than MOPS, making the MES SDS Running Buffer faster than the MOPS SDS running buffers. The difference in ion migration affects stacking and results in a difference in protein separation range between these buffers. Use of MOPS buffer allows proteins to run slower than when using MES buffer.

Compare protein migration patterns using MES and MOPs on Bolt Bis-Tris Plus gels

See all available buffers and reagents available for SDS-PAGE

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name or Material
Running Buffer
Recommended Storage
MOPS SDS Running Buffer (20X).

Store at 4°C to 25°C.

Shelf-life is 6 months, when stored properly.
Concentration
20X
For Use With (Equipment)
Bolt
Physical Form
Liquid
Product Line
Bolt
Quantity
5 L

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text