Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Thermo Scientific™

Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems

Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.
Have Questions?
Catalog NumberIncludes
B1-BPBuffer Exchange Ports
Catalog number B1-BP
Price (EUR)
-
Request A Quote
Includes:
Buffer Exchange Ports
Ask our AI about this Product

Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.

Features:

  • System offers a wide range of combs.
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.

Includes:

  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
  • Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick

Ordering Information:

Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.

Warranty and Service Offering:

  • Three years
Specifications
Certifications/ComplianceCE marked
Dimensions (L x W x H)8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm)
Height (English)3.75 in.
Height (Metric)9.5 cm
IncludesBuffer Exchange Ports
Length (English)8.63 in.
Length (Metric)22 cm
Length (Metric) Gel11 cm
Sample Size5 to 34
Volume (Metric) Buffer600 mL
Width (English)5.88 in.
Width (Metric)15 cm
Width (Metric) Gel9 cm
Gel SizeMini
Product LineOwl
TypeMini Gel Electrophoresis System
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

Yes, it is compatible with the Owl Gel Systems. The following ones can be used with the PowerEase power supplies with no need for adaptors:

    Owl EasyCast B1 Mini Gel Electrophor. Sys. (Cat. No. B1-BP)
    Owl A2 Large Gel Sys. (Cat. No. A2)
    Owl EasyCast B3 Gel Sys. with Built-In Recirculat. (Cat. No. B3).


Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.

Share catalog number, name or link

1x1 image pixel for data collection