Thermo Scientific Chemicals

Periodic acid, 99%, Thermo Scientific Chemicals

Catalog number: B20433.0B
1000 g, Each
Thermo Scientific Chemicals

Periodic acid, 99%, Thermo Scientific Chemicals

Catalog number: B20433.0B
1000 g, Each
Quantity
Catalog number: B20433.0B
also known as B20433-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10450-60-9
IUPAC Name
tetrahydroxyiodous acid
Molecular Formula
H5IO6
InChI Key
TWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILES
O[I](O)(O)(O)(O)=O
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystalline or lumpy powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Iodometric Titration)
≥98.5 to ≤101.5%

Description

Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.

Solubility
Soluble in water and alcohols.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, strong bases and dimethyl sulfoxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text