Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

BamHI-Lsp1109I Buffer (10X)

Catalog number: B57
Have Questions?
Thermo Scientific™

BamHI-Lsp1109I Buffer (10X)

Catalog number: B57
Have Questions?
Catalog Number
B57
Unit Size
5 x 1.0 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
B575 x 1.0 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific 10X Buffer BamHI, Lsp1109I is the optimal buffer recommended for use with BamHI and Lsp1109I restriction enzymes and is premixed with BSA for enhanced stability.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

Thermo Scientific restriction enzymes exhibit 100% of their certified activity in the recommended buffer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
10X
Product Type
BamHI-Lsp1109I Buffer
Research Category
Traditional Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text