Thermo Scientific™ 10X DreamTaq™ Buffer is specifically optimized for PCR using DreamTaq™ DNA Polymerase and DreamTaq™ Hot Start DNA Polymerase. The buffer contains both KCl and (NH4)2SO4 to provide high specificity of primer annealing at a wide range of MgCl2 concentrations. 10X DreamTaq buffer includes 20 mM MgCl2.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Polymerase
DreamTaq Hot Start DNA Polymerase, DreamTaq DNA Polymerase
Product Line
DreamTaq
Concentration
10X
Volume
1.25 mL
Contents & Storage
Contains: • 4 x 1.25 mL 10X DreamTaq Buffer (includes 20 mM MgCl2)