Thermo Scientific™
Open Top Screw Caps for Sample Storage Vials
Catalog number: B7807-20
Have Questions?
Thermo Scientific™
Open Top Screw Caps for Sample Storage Vials
Catalog number: B7807-20
Have Questions?
Open Top Screw Caps for Sample Storage Vials
Catalog Number
B7807-20
Unit Size
Pack of 100
dimClosureSize
Price (USD)
Full specifications
Quantity
Closure ColorBlack
DescriptionOpen top 20-400 Screw Cap
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open-Top)
Quantity100
Closure Size20 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
dimClosureSize
Price (USD)
Full specifications
Quantity
Closure ColorWhite
DescriptionOpen top 24-400 Screw Cap
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open-Top)
Quantity100
Closure Size24 mm
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
dimClosureSize
Price (USD)
Full specifications
Quantity
Closure ColorBlack
DescriptionOpen top 15-425 Screw Cap
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open-Top)
Quantity100
Closure Size15 mm
Unit SizePack of 100
Showing 3 of 3
Catalog NumberSpecificationsUnit SizedimClosureSizePrice (USD)Quantity
B7807-20Full specifications
Pack of 100
Closure ColorBlack
DescriptionOpen top 20-400 Screw Cap
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open-Top)
Quantity100
Closure Size20 mm
Unit SizePack of 100
B7807-24Full specifications
Pack of 100
Closure ColorWhite
DescriptionOpen top 24-400 Screw Cap
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open-Top)
Quantity100
Closure Size24 mm
Unit SizePack of 100
B7807-15Full specifications
Pack of 100
Closure ColorBlack
DescriptionOpen top 15-425 Screw Cap
Closure MaterialPolypropylene
Closure TypeScrew Cap (Open-Top)
Quantity100
Closure Size15 mm
Unit SizePack of 100
Showing 3 of 3
Open Top Screw Caps for Sample Storage Vials