Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Thermo Scientific™
Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Get improved plasma stability from the tulip design and easy, push-in insertion of Thermo Scientific™ torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Have Questions?
Change view
Catalog Number
Type
BRE0024760
ICP-MS PLUS Torch
1230790
ICP-MS Quartz Torch
BRE0013019
ICP-MS Organic Torch
3 Options
Catalog number BRE0024760
Price (USD)
5,430.00
Each
Add to cart
Type:
ICP-MS PLUS Torch
Recurring order eligible. Learn more »
Price (USD)
5,430.00
Each
Add to cart
Ask our AI about this Product
Gain improved plasma stability from the tulip design and easy, push-in Gain improved plasma stability from the tulip design and easy, push-in insertion of fully demountable, single piece, Thermo Scientific™ Torches with connection-free gas supply for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems. All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
Specifications
For Use With (Equipment)iCAP MX and iCAP Q/Qnova Series ICP-MS Systems
TypeICP-MS PLUS Torch
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.